Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • GRASP65 Antibodies

Invitrogen

GRASP65 Recombinant Rabbit Monoclonal Antibody (JB36-34)

View all (18) GRASP65 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite GRASP65 Recombinant Rabbit Monoclonal Antibody (JB36-34)

  • Antibody Testing Data (7)
GRASP65 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
GRASP65 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 7

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

GRASP65 Antibody (MA5-34658) in ICC/IF

Immunofluorescent analysis of GRASP65 in MCF-7 cells (green). Samples were fixed in paraformaldehyde and permeabilised with 0.25% Triton X100/PBS, incubated with GRASP65 monoclonal antibody (Product # MA5-34658), followed by DAPI (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

GRASP65 Antibody in Immunocytochemistry (ICC/IF)
GRASP65 Antibody in Immunocytochemistry (ICC/IF)
GRASP65 Antibody in Immunocytochemistry (ICC/IF)
GRASP65 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
GRASP65 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
GRASP65 Antibody in Western Blot (WB)
GRASP65 Antibody in Flow Cytometry (Flow)
GRASP65 Recombinant Rabbit Monoclonal Antibody (JB36-34)

Product Details

MA5-34658

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-

Immunocytochemistry (ICC/IF)

1:50-1:200
-

Flow Cytometry (Flow)

1:50-1:100
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

JB36-34

Immunogen

Synthetic peptide within Human GRASP65 aa 396-440
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

TBS, pH 7.4, with 40% glycerol, 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2848566

Product Specific Information

Positive Control: THP-1,SKBR3, HUVEC, HepG2, MCF-7, human colon cancer tissue, human uterine tissue.

Target Information

Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Golgi peripheral membrane protein p65; Golgi phosphoprotein 5; Golgi reassembly and stacking protein 1; golgi reassembly stacking protein 1, 65kDa; Golgi reassembly-stacking protein 1; Golgi reassembly-stacking protein of 65 kDa; GOLPH5; GRASP65

View more View less

Gene Aliases: GOLPH5; GORASP1; GRASP65; P65

View more View less

UniProt ID: (Human) Q9BQQ3

View more View less

Entrez Gene ID: (Human) 64689

View more View less

Function(s)
protein binding
Process(es)
protein N-linked glycosylation ER to Golgi vesicle-mediated transport Golgi organization protein transport COPII vesicle coating negative regulation of dendrite morphogenesis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0