Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • GNAO1 Antibodies

Invitrogen

GNAO1 Polyclonal Antibody

View all (12) GNAO1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite GNAO1 Polyclonal Antibody

  • Antibody Testing Data (1)
GNAO1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
GNAO1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

GNAO1 Antibody (PA5-97796) in WB

Western Blot analysis of GNAO1 using a GNAO1 Polyclonal antibody (Product # PA5-97796) at a concentration of 3 µg/mL. Positive WB detected in: Rat brain tissue, Mouse brain tissue. A secondary Goat polyclonal antibody to rabbit IgG was applied at a 1:50,000 dilution. Observed band size: 41 kDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

GNAO1 Antibody in Western Blot (WB)
GNAO1 Polyclonal Antibody

Product Details

PA5-97796

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:5,000
-

ELISA (ELISA)

Assay-Dependent
-
Product Specifications

Species Reactivity

Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant Mouse Guanine nucleotide-binding protein G(o) subunit alpha protein (2-354AA)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

4 mg/mL

Purification

Protein G

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.03% ProClin 300

Storage conditions

-20°C or -80°C if preferred

Shipping conditions

Wet ice

RRID

AB_2812411

Target Information

Heterotrimeric G proteins are membrane bound GTPases that are linked to 7-TM receptors. Each G protein contains an alpha-, beta- and gamma-subunit and is bound to GDP in the 'off' state. Ligand-receptor binding results in detachment of the G protein, switching it to an 'on' state and permitting Galpha activation of second messenger signalling cascades. There are several types of Galpha proteins; Galphas and Galphai bind directly to adenylyl cyclase and stimulate or inhibit its activity, leading to an increase or decrease in cAMP levels respectively.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: DKFZp686O0962; G alpha o; GNAO 1; Go alpha; GTP-binding protein alpha o; GTP-binding protein alpha-o subunit; guanine nucleotide binding protein, alpha O polypeptide 1; Guanine nucleotide-binding protein G(o) subunit alpha

View more View less

Gene Aliases: alphaO; AW050213; Galphao; Gna0; Gnao; Gnao1; RATBPGTPC

View more View less

UniProt ID: (Rat) P59215, (Mouse) P18872

View more View less

Entrez Gene ID: (Rat) 50664, (Mouse) 14681

View more View less

Function(s)
GTPase activity signal transducer activity protein binding GTP binding GDP binding G-protein beta/gamma-subunit complex binding G-protein coupled serotonin receptor binding mu-type opioid receptor binding protein complex binding GTPase activating protein binding metal ion binding corticotropin-releasing hormone receptor 1 binding nucleotide binding guanyl nucleotide binding
Process(es)
G-protein coupled receptor signaling pathway adenylate cyclase-modulating G-protein coupled receptor signaling pathway dopamine receptor signaling pathway aging locomotory behavior regulation of heart contraction metabolic process cellular process response to organonitrogen compound response to organic cyclic compound forebrain development neuron projection development response to cytokine response to drug response to hydrogen peroxide response to morphine positive regulation of GTPase activity negative regulation of calcium ion transport signal transduction
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0