Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • GLUT5 Antibodies

Invitrogen

GLUT5 Polyclonal Antibody

3 Published Figures
3 References
View all (38) GLUT5 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite GLUT5 Polyclonal Antibody

  • Antibody Testing Data (6)
  • Published Figures (3)
GLUT5 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
GLUT5 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 9

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

GLUT5 Antibody (PA5-80023) in ICC/IF

Immunocytochemistry analysis of SLC2A5 using anti-SLC2A5 antibody (Product # PA5-80023) . SLC2A5 was detected in a section of U2OS cells. Enzyme antigen retrieval was performed using IHC enzyme antigen retrieval reagent for 15 mins. The cells were blocked with 10% goat serum and then incubated with 2μg/mL rabbit anti-SLC2A5 antibody (Product # PA5-80023) overnight at 4°C. DyLight®488 Conjugated Goat Anti-Rabbit IgG was used as secondary antibody at 1:100 dilution and incubated for 30 minutes at 37°C. The section was counterstained with DAP... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

GLUT5 Antibody in Immunocytochemistry (ICC/IF)
GLUT5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
GLUT5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
GLUT5 Antibody in Western Blot (WB)
GLUT5 Antibody in Flow Cytometry (Flow)
GLUT5 Antibody in Flow Cytometry (Flow)
GLUT5 Antibody in Western Blot (WB)
GLUT5 Antibody in Immunohistochemistry (IHC)
GLUT5 Antibody in Western Blot (WB)
GLUT5 Polyclonal Antibody

Product Details

PA5-80023

Applications
Tested Dilution
Publications

Western Blot (WB)

0.1-0.5 µg/mL
View 2 publications 2 publications

Immunohistochemistry (IHC)

-
View 1 publication 1 publication

Immunohistochemistry (Paraffin) (IHC (P))

0.5-1 µg/mL
-

Immunohistochemistry (Frozen) (IHC (F))

0.5-1 µg/mL
-

Immunocytochemistry (ICC/IF)

2 µg/mL
-

Flow Cytometry (Flow)

1-3 µg/1x10^6 cells
-
Product Specifications

Species Reactivity

Human, Rat

Published species

Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

A synthetic peptide corresponding to a sequence in the middle region of human SLC2A5 (223-259aa QKKDEAAAKKALQTLRGWDSVDREVAEIRQEDEAEKA).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

500 µg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS with 5mg BSA

Contains

0.05mg sodium azide

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2747138

Product Specific Information

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.

Positive Control - WB: rat brain tissue, K562 whole cell. IHC: rat intestinal cancer tissue, human intestinal cancer tissue. ICC/IF: U20S cell. Flow: THP-1 cell.

Target Information

Cytochalasin B-sensitive carrier. Seems to function primarily as a fructose transporter.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: Fructose transporter; Glucose transporter type 5, small intestine; glucose transporter-like protein 5; GLUT-5; RP5-963K15.1; solute carrier family 2 (facilitated glucose transporter), member 5; solute carrier family 2 (facilitated glucose/fructose transporter), member 5; Solute carrier family 2, facilitated glucose transporter member 5; solute carrier family 2, member 5; testicular tissue protein Li 81

View more View less

Gene Aliases: GLUT-5; GLUT5; SLC2A5

View more View less

UniProt ID: (Human) P22732, (Rat) P43427

View more View less

Entrez Gene ID: (Human) 6518, (Rat) 65197

View more View less

Function(s)
fructose transmembrane transporter activity glucose transmembrane transporter activity fructose binding
Process(es)
regulation of systemic arterial blood pressure mediated by a chemical signal carbohydrate metabolic process hexose transport response to fructose fructose transport glucose transport cellular response to fructose stimulus glucose transmembrane transport fructose import across plasma membrane transmembrane transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0