Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Connexin 32 Antibodies

Abnova

GJB1 Monoclonal Antibody (2B9)

View all (22) Connexin 32 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite GJB1 Monoclonal Antibody (2B9)

  • Antibody Testing Data (1)
GJB1 Antibody in ELISA (ELISA)
Group 53 Created with Sketch.
GJB1 Antibody in ELISA (ELISA)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

GJB1 Antibody (H00002705-M03) in ELISA

Detection limit for recombinant GST tagged GJB1 is 0.3 ng/mL as a capture antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

GJB1 Antibody in ELISA (ELISA)
GJB1 Monoclonal Antibody (2B9)

Product Details

H00002705-M03

Applications
Tested Dilution
Publications

ELISA (ELISA)

0.3 ng/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

2B9

Immunogen

GJB1 (AAH22426, 75 a.a. approximately 174 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Sequence of this protein is as follows: RLWSLQLILV STPALLVAMH VAHQQHIEKK MLRLEGHGDP LHLEEVKRHK VHISGTLWWT YVISVVFRLL FEAVFMYVFY LLYPGYAMVR LVKCDVYPCP

Target Information

This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: connexin 32; Connexin-32; Cx32; GAP junction 28 kDa liver protein; Gap junction beta-1 protein; gap junction protein, beta 1, 32kDa

View more View less

Gene Aliases: CMTX; CMTX1; CX32; GJB1

View more View less

UniProt ID: (Human) P08034

View more View less

Entrez Gene ID: (Human) 2705

View more View less

Function(s)
gap junction channel activity protein homodimerization activity
Process(es)
transport cell-cell signaling nervous system development purine ribonucleotide transport gap junction assembly protein oligomerization transmembrane transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0