Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • FZD7 Antibodies

Proteintech

Frizzled 7 Polyclonal Antibody

12 References
View all (12) FZD7 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Frizzled 7 Polyclonal Antibody

  • Antibody Testing Data (4)
Frizzled 7 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
Frizzled 7 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Frizzled 7 Antibody (16974-1-AP) in WB

Mouse skeletal muscle tissue were subjected to SDS PAGE followed by western blot with 16974-1-AP (Frizzled 7 antibody) at dilution of 1:600 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Frizzled 7 Antibody in Western Blot (WB)
Frizzled 7 Antibody in Western Blot (WB)
Frizzled 7 Antibody in Western Blot (WB)
Frizzled 7 Antibody in Immunoprecipitation (IP)
Frizzled 7 Polyclonal Antibody

Product Details

16974-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
View 9 publications 9 publications

Immunohistochemistry (IHC)

-
View 1 publication 1 publication

Immunoprecipitation (IP)

0.5-4.0 µg
View 1 publication 1 publication

Not applicable (N/A)

-
View 1 publication 1 publication
Product Specifications

Species Reactivity

Human, Mouse

Published species

Human, Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Frizzled 7 Fusion Protein Ag9446 (33-257 aa encoded by BC015915)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.33 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: QPYHGEKGI SVPDHGFCQP ISIPLCTDIA YNQTILPNLL GHTNQEDAGL EVHQFYPLVK VQCSPELRFF LCSMYAPVCT VLDQAIPPCR SLCERARQGC EALMNKFGFQ WPERLRCENF PVHGAGEICV GQNTSDGSGG PGGGPTAYPT APYLPDLPFT ALPPGASDGR GRPAFPFSCP RQLKVPPYLG YRFLGERDCG APCEPGRANG LMYFKEEERR FARLWV (33-257 aa encoded by BC015915)

Target Information

Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD7 protein contains an N-terminal signal sequence, 10 cysteine residues typical of the cysteine-rich extracellular domain of Fz family members, 7 putative transmembrane domains, and an intracellular C-terminal tail with a PDZ domain-binding motif. FZD7 gene expression may downregulate APC function and enhance beta-catenin-mediated signals in poorly differentiated human esophageal carcinomas.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: frizzled 7, seven transmembrane spanning receptor; frizzled family receptor 7; frizzled homolog 7; Frizzled, drosophila, homolog of, 7; Frizzled-7; Fz-7; FzE3; hFz7; mFz7; rizzled 7

View more View less

Gene Aliases: Fz7; FZD7; FzE3

View more View less

UniProt ID: (Human) O75084, (Mouse) Q61090

View more View less

Entrez Gene ID: (Human) 8324, (Mouse) 14369

View more View less

Function(s)
G-protein coupled receptor activity frizzled binding protein binding Wnt-protein binding PDZ domain binding Wnt-activated receptor activity signal transducer activity transmembrane signaling receptor activity
Process(es)
regulation of transcription, DNA-templated signal transduction cell surface receptor signaling pathway G-protein coupled receptor signaling pathway multicellular organism development negative regulation of cell-substrate adhesion skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration Wnt signaling pathway stem cell population maintenance T cell differentiation in thymus substrate adhesion-dependent cell spreading regulation of catenin import into nucleus non-canonical Wnt signaling pathway non-canonical Wnt signaling pathway via JNK cascade positive regulation of phosphorylation negative regulation of ectodermal cell fate specification positive regulation of transcription, DNA-templated somatic stem cell division positive regulation of epithelial cell proliferation involved in wound healing canonical Wnt signaling pathway mesenchymal to epithelial transition neuron differentiation positive regulation of JNK cascade Wnt signaling pathway, planar cell polarity pathway cellular response to retinoic acid
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0