Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Ferroportin Antibodies

Invitrogen

Ferroportin Polyclonal Antibody

View all (11) Ferroportin antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Ferroportin Polyclonal Antibody

  • Antibody Testing Data (2)
Ferroportin Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
Ferroportin Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Ferroportin Antibody (PA5-143047) in ICC/IF

Immunocytochemistry-Immunofluorescence analysis of Ferroportin in HeLa cells using Ferroportin Polyclonal Antibody (Product # PA5-143047). Cells were fixed with paraformaldehyde, permeabilized with 0.15% Triton, and incubated with the primary antibody for 1hr (10 µg/mL) followed by Alexa Fluor 488 secondary antibody (2 µg/mL), showing cytoplasmic and plasma membrane staining. The nuclear stain is DAPI (blue... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Ferroportin Antibody in Immunocytochemistry (ICC/IF)
Ferroportin Antibody in Flow Cytometry (Flow)
Ferroportin Polyclonal Antibody

Product Details

PA5-143047

Applications
Tested Dilution
Publications

Western Blot (WB)

1-2 µg/mL
-

Immunocytochemistry (ICC/IF)

10 µg/mL
-

Flow Cytometry (Flow)

10 µg/mL
- -
Product Specifications

Species Reactivity

Human

Host/Isotype

Goat / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Peptide with sequence C-ELKQLNLHKDTEPKP (aa 245-259).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.5 mg/mL

Purification

Ammonium sulfate precipitation

Storage buffer

TBS, pH 7.3, with 0.5% BSA

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2933686

Product Specific Information

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

Target Information

SLC40A1 is an iron-regulated transporter assumed to play roles in intestinal iron absorption and cellular iron release. It mediates iron efflux in the presence of ferroxidase. Defects in SLC40A1 cause hemochromatosis type 4 (HFE4), which is an autosomal dominant iron-loading disorder.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Ferroportin; Ferroportin-1; iron regulated gene 1; Iron-regulated transporter 1; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3; solute carrier family 40 (iron-regulated transporter), member 1; Solute carrier family 40 member 1

View more View less

Gene Aliases: FPN; FPN1; HFE4; IREG1; MST079; MSTP079; MTP1; SLC11A3; SLC40A1

View more View less

UniProt ID: (Human) Q9NP59

View more View less

Entrez Gene ID: (Human) 30061

View more View less

Function(s)
receptor activity iron ion transmembrane transporter activity protein binding ferrous iron transmembrane transporter activity peptide hormone binding iron channel activity
Process(es)
lymphocyte homeostasis endothelium development cellular iron ion homeostasis regulation of transcription from RNA polymerase II promoter in response to iron iron ion transmembrane transport negative regulation of apoptotic process positive regulation of transcription from RNA polymerase II promoter spleen trabecula formation multicellular organismal iron ion homeostasis ferrous iron export
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-657bd577ff-sg76p:80/100.66.128.142:80.
git-commit: cdba60da2f3289684898c31dc50804c008488b19
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0