Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • FAM20A Antibodies

Invitrogen

FAM20A Recombinant Rabbit Monoclonal Antibody (2I9B0)

View all (5) FAM20A antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite FAM20A Recombinant Rabbit Monoclonal Antibody (2I9B0)

  • Antibody Testing Data (1)
FAM20A Antibody in Western Blot (WB)
Group 53 Created with Sketch.
FAM20A Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

FAM20A Antibody (MA5-55443) in WB

Western Blot analysis of various lysates using FAM20A Recombinant Monoclonal Antibody (Product # MA5-55443) at 1:20,000 dilution. Secondary antibody: HRP-conjugated Goat anti-Rabbit IgG (H+L) at 1:10,000 dilution. Lysates/proteins: 25 μg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Enhanced Kit. Exposure time: 180s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

FAM20A Antibody in Western Blot (WB)
FAM20A Recombinant Rabbit Monoclonal Antibody (2I9B0)

Product Details

MA5-55443

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000-1:20,000
-

ELISA (ELISA)

1 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

2I9B0

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 400-500 of human FAM20A.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.86 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.3, with 0.05% BSA, 50% glycerol

Contains

0.05% ProClin 300

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

RRID

AB_3667684

Product Specific Information

Immunogen Sequence: AIFDFLIGNM DRHHYEMFTK FGDDGFLIHL DNARGFGRHS HDEISILSPL SQCCMIKKKT LLHLQLLAQA DYRLSDVMRE SLLEDQLSPV LTEPHLLALD R

Target Information

This locus encodes a protein that is likely secreted and may function in hematopoiesis. A mutation at this locus has been associated with amelogenesis imperfecta and gingival hyperplasia syndrome. Alternatively spliced transcript variants have been identified.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: family with sequence similarity 20, member A; protein FAM20A; Pseudokinase FAM20A

View more View less

Gene Aliases: AI1G; AIGFS; FAM20A; FP2747; UNQ9388/PRO34279

View more View less

UniProt ID: (Human) Q96MK3

View more View less

Entrez Gene ID: (Human) 54757

View more View less

Function(s)
protein serine/threonine kinase activity protein binding protein serine/threonine kinase activator activity
Process(es)
positive regulation of protein phosphorylation biomineral tissue development tooth eruption calcium ion homeostasis enamel mineralization positive regulation of protein serine/threonine kinase activity
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0