Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • EphA2 Antibodies

Abnova

EPHA2 Monoclonal Antibody (1E3)

View all (44) EphA2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite EPHA2 Monoclonal Antibody (1E3)

  • Antibody Testing Data (4)
EPHA2 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
EPHA2 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

EPHA2 Antibody (H00001969-M02) in ICC/IF

Immunofluorescence of monoclonal antibody to EPHA2 on A-431 cell. Antibody concentration is 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

EPHA2 Antibody in Immunocytochemistry (ICC/IF)
EPHA2 Antibody in Western Blot (WB)
EPHA2 Antibody in Western Blot (WB)
EPHA2 Antibody in Western Blot (WB)
EPHA2 Monoclonal Antibody (1E3)

Product Details

H00001969-M02

Applications
Tested Dilution
Publications

Western Blot (WB)

1-5 µg/mL
-

Immunocytochemistry (ICC/IF)

10 µg/mL
-

ELISA (ELISA)

10 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG3, kappa

Class

Monoclonal

Type

Antibody

Clone

1E3

Immunogen

EPHA2 (AAH37166, 204 a.a. approximately 326 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Sequence of this protein is as follows: LLQGLAHFPE TIAGSDAPSL ATVAGTCVDH AVVPPGGEEP RMHCAVDGEW LVPIGQCLCQ AGYEKVEDAC QACSPGFFKF EASESPCLEC PEHTLPSPEG ATSCECEEGF FRAPQDPASM PCT

Target Information

EPHA2 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA2 is overexpressed in many types of human cancers. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e.g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e.g. ABL, FAK, JAK, and SRC families).

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Ephrin type-A receptor 2; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; soluble EPHA2 variant 1; Tyrosine-protein kinase receptor ECK

View more View less

Gene Aliases: ARCC2; CTPA; CTPP1; CTRCT6; ECK; EPHA2

View more View less

UniProt ID: (Human) P29317

View more View less

Entrez Gene ID: (Human) 1969

View more View less

Function(s)
transmembrane receptor protein tyrosine kinase activity ephrin receptor activity protein binding ATP binding cadherin binding involved in cell-cell adhesion
Process(es)
skeletal system development angiogenesis vasculogenesis osteoblast differentiation multicellular organism development intrinsic apoptotic signaling pathway in response to DNA damage regulation of lamellipodium assembly notochord formation viral process cell migration peptidyl-tyrosine phosphorylation neural tube development neuron differentiation keratinocyte differentiation osteoclast differentiation mammary gland epithelial cell proliferation regulation of cell adhesion mediated by integrin post-anal tail morphogenesis protein kinase B signaling regulation of blood vessel endothelial cell migration regulation of angiogenesis bone remodeling ephrin receptor signaling pathway axial mesoderm formation negative regulation of protein kinase B signaling notochord cell development cell chemotaxis branching involved in mammary gland duct morphogenesis lens fiber cell morphogenesis regulation of ERK1 and ERK2 cascade response to growth factor positive regulation of establishment of protein localization to plasma membrane activation of GTPase activity cell-cell adhesion
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0