Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • EpCAM (CD326) Antibodies

Proteintech

EPCAM Monoclonal Antibody (2A2D5)

6 References
View all (272) EpCAM (CD326) antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite EPCAM Monoclonal Antibody (2A2D5)

  • Antibody Testing Data (11)
EPCAM Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
EPCAM Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 11

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

EPCAM Antibody (66316-1-IG) in ICC/IF

Immunofluorescent analysis of (-20°C Methanol) fixed MCF-7 cells using EPCAM antibody (Product # 66316-1-Ig) (Clone: 2A2D5) at dilution of 1:400 and CoraLite®488-Conjugated AffiniPure Goat Anti-Mouse IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

EPCAM Antibody in Immunocytochemistry (ICC/IF)
EPCAM Antibody in Immunocytochemistry (ICC/IF)
EPCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
EPCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
EPCAM Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
EPCAM Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
EPCAM Antibody in Western Blot (WB)
EPCAM Antibody in Western Blot (WB)
EPCAM Antibody in Western Blot (WB)
EPCAM Antibody in Western Blot (WB)
EPCAM Antibody in Flow Cytometry (Flow)
EPCAM Monoclonal Antibody (2A2D5)

Product Details

66316-1-IG

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000-1:4,000
View 4 publications 4 publications

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:500
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1:50-1:500
-

Immunocytochemistry (ICC/IF)

1:50-1:500
-

Flow Cytometry (Flow)

Assay-dependent
View 1 publication 1 publication

Not applicable (N/A)

-
View 1 publication 1 publication
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2a

Class

Monoclonal

Type

Antibody

Clone

2A2D5

Immunogen

EPCAM Fusion Protein Ag15393 (24-264 aa encoded by BC014785)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated
  • CoraLite 594
  • CoraLite Plus 488
  • Request custom conjugation

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: QEECVCENY KLAVNCFVNN NRQCQCTSVG AQNTVICSKL AAKCLVMKAE MNGSKLGRRA KPEGALQNND GLYDPDCDES GLFKAKQCNG TSTCWCVNTA GVRRTDKDTE ITCSERVRTY WIIIELKHKA REKPYDSKSL RTALQKEITT RYQLDPKFIT SILYENNVIT IDLVQNSSQK TQNDVDIADV AYYFEKDVKG ESLFHSKKMD LTVNGEQLDL DPGQTLIYYV DEKAPEFSMQ GL (24-264 aa encoded by BC014785)

Target Information

Ep-CAM (epithelial adhesion molecule, epithelial specific antigen, ESA) is a transmembrane glycoprotein expressed in the epithelium with a molecular weight of approximately 40 kDa, which functions as an epithelial cell adhesion molecule. Ep-CAM functions as a homotypic calcium-independent cell adhesion molecule, and has a direct impact on cell cycle, proliferation and metabolism of epithelial cells and fibroblasts due to its ability to rapidly induce the proto-oncogene c-myc and the cell cycle regulating genes cyclin A and E. Ep-CAM mediates Ca2+-independent homotypic interactions. Formation of Ep-CAM-mediated adhesions have a negative regulatory effect on adhesions mediated by classic cadherins, which may have strong effects on the differentiation and growth of epithelial cells. Ep-CAM overexpression was suggested to be associated with enhanced epithelial proliferation. Ep-CAM is highly expressed in human carcinomas, and is a marker for tumors of epithelial lineage. Ep-CAM is expressed on baso-lateral cell surface in most simple epithelia and many carcinoma types. Also, Ep-CAM reportedly distinguishes adenocarcinomas from pleural mesotheliomas.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: Adenocarcinoma-associated antigen; CD326; Cell surface glycoprotein Trop-1; EGP; EGP314; Ep-CAM; Epithelial cell adhesion molecule; Epithelial cell surface antigen; Epithelial glycoprotein; Epithelial glycoprotein 314; hEGP314; human epithelial glycoprotein-2; KS 1/4 antigen; KSA; Major gastrointestinal tumor-associated protein GA733-2; membrane component, chromosome 4, surface marker (35kD glycoprotein); Tumor-associated calcium signal transducer 1

View more View less

Gene Aliases: DIAR5; EGP-2; EGP314; EGP40; EPCAM; ESA; GA733-2; HNPCC8; KS1/4; KSA; M1S2; M4S1; MIC18; MK-1; TACSTD1; TROP1

View more View less

UniProt ID: (Human) P16422

View more View less

Entrez Gene ID: (Human) 4072

View more View less

Function(s)
protein binding protein complex binding cadherin binding involved in cell-cell adhesion
Process(es)
ureteric bud development positive regulation of cell proliferation signal transduction involved in regulation of gene expression negative regulation of apoptotic process positive regulation of transcription from RNA polymerase II promoter stem cell differentiation cell-cell adhesion negative regulation of cell-cell adhesion mediated by cadherin positive regulation of cell motility positive regulation of stem cell proliferation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0