Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • PHD1 Antibodies

OriGene

EGLN2 Monoclonal Antibody (OTI5E11), TrueMAB™

View all (15) PHD1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite EGLN2 Monoclonal Antibody (OTI5E11), TrueMAB™

  • Antibody Testing Data (1)
EGLN2 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
EGLN2 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

EGLN2 Antibody (TA808685) in WB

HEK293T cells were transfected with the pCMV6-ENTRY control (Left lane) or pCMV6-ENTRY EGLN2 (RC219191, Right lane) cDNA for 48 hrs and lysed. Equivalent amounts of cell lysates (5 µg per lane) were separated by SDS-PAGE and immunoblotted with anti-EGLN2. Positive lysates LY409048 (100 µg) and LC409048 (20 µg) can be purchased separately from OriGene. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

EGLN2 Antibody in Western Blot (WB)
EGLN2 Monoclonal Antibody (OTI5E11), TrueMAB™

Product Details

TA808685

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI5E11

Immunogen

Full length human recombinant protein of human EGLN2 produced in E.coli.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 1% BSA, 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

Prolyl hydroxylase domain proteins HIF PHD1, HIF PHD2 and HIF PHD3 (known as PHD1, PHD2 and PHD3 in rodents, respectively) can hydroxylate HIF-alpha subunits. Hypoxia-inducible factor (HIF) is a transcriptional regulator important in several aspects of oxygen homeostasis. The prolyl hydroxylases catalyze the posttranslational formation of 4-hydroxyproline in HIF-alpha proteins. HIF PHD1, which is widely expressed, with highest levels of expression in testis, functions as a cellular oxygen sensor and is important in cell growth regulation. HIF PHD1 can localize to the nucleus or the cytoplasm and is also detected in hormone responsive tissues, such as normal and cancerous mammary, ovarian and prostate epithelium. HIF PHD1 is encoded by EGLN2, which maps to chromosome 19q13. 3.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Egl nine homolog 2; egl-9 family hypoxia-inducible factor 2; EIT-6; Estrogen-induced tag 6; HIF-P4H 1; HIF-PH1; HIF-prolyl hydroxylase 1; HPH-3; Hypoxia-inducible factor prolyl hydroxylase 1; P4H1; PHD1; Prolyl hydroxylase domain-containing protein 1; Prolyl hydroxylase EGLN2

View more View less

Gene Aliases: EGLN2; EIT6; HIF-PH1; HIFPH1; HPH-1; HPH-3; PHD1

View more View less

UniProt ID: (Human) Q96KS0

View more View less

Entrez Gene ID: (Human) 112398

View more View less

Function(s)
protein binding ferrous iron binding oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors oxygen sensor activity L-ascorbic acid binding peptidyl-proline 4-dioxygenase activity
Process(es)
regulation of cell growth response to hypoxia peptidyl-proline hydroxylation to 4-hydroxy-L-proline intracellular estrogen receptor signaling pathway regulation of neuron apoptotic process cell redox homeostasis positive regulation of protein catabolic process oxidation-reduction process regulation of transcription from RNA polymerase II promoter in response to hypoxia
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0