Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Dopamine beta Hydroxylase Antibodies

Invitrogen

Dopamine beta Hydroxylase Polyclonal Antibody

View all (25) Dopamine beta Hydroxylase antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Dopamine beta Hydroxylase Polyclonal Antibody

  • Antibody Testing Data (1)
Dopamine beta Hydroxylase Antibody in Western Blot (WB)
Group 53 Created with Sketch.
Dopamine beta Hydroxylase Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Dopamine beta Hydroxylase Antibody (PA5-86705) in WB

Western blot analysis of Dopamine beta Hydroxylase in Lane 1: HEK293T cell lysate, Lane 2: sp2/0 cell lysate, Lane 3: NIH-3T3 cell lysate. Sample was incubated with Dopamine beta Hydroxylase polyclonal antibody (Product # PA5-86705) using a 1:500 dilution. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Dopamine beta Hydroxylase Antibody in Western Blot (WB)
Dopamine beta Hydroxylase Polyclonal Antibody

Product Details

PA5-86705

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Synthetic peptide, corresponding to amino acids 445-490 of Human DBH.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.2, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Wet ice

RRID

AB_2803465

Product Specific Information

Purity is > 95% by SDS-PAGE.

Target Information

The protein encoded by this gene is an oxidoreductase belonging to the copper type II, ascorbate-dependent monooxygenase family. It is present in the synaptic vesicles of postganglionic sympathetic neurons and converts dopamine to norepinephrine. It exists in both soluble and membrane-bound forms, depending on the absence or presence, respectively, of a signal peptide.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Dopamine beta hydroxylase (dopamine beta-monooxygenase); Dopamine beta-hydroxylase; dopamine beta-hydroxylase (dopamine beta-monooxygenase); Dopamine beta-monooxygenase; Dopamine-Beta-hydroxylase

View more View less

Gene Aliases: DBH; DBM; DOPBHY

View more View less

UniProt ID: (Human) P09172, (Rat) Q05754, (Mouse) Q64237

View more View less

Entrez Gene ID: (Human) 1621, (Rat) 25699, (Mouse) 13166

View more View less

Function(s)
catalytic activity dopamine beta-monooxygenase activity copper ion binding L-ascorbic acid binding oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced ascorbate as one donor, and incorporation of one atom of oxygen monooxygenase activity oxidoreductase activity metal ion binding
Process(es)
cytokine production blood vessel remodeling response to amphetamine leukocyte mediated immunity catecholamine metabolic process memory locomotory behavior associative learning visual learning response to iron ion response to ozone multicellular organism aging response to organic cyclic compound response to estradiol social behavior response to isolation stress response to immobilization stress regulation of cell proliferation homoiothermy dopamine catabolic process norepinephrine biosynthetic process response to drug glucose homeostasis fear response maternal behavior response to peptide hormone response to ethanol positive regulation of vasoconstriction octopamine metabolic process response to copper ion behavioral response to ethanol response to pain leukocyte migration sensory perception of taste oxidation-reduction process bone development cellular response to manganese ion cellular response to nicotine regulation of extrinsic apoptotic signaling pathway chemical synaptic transmission catecholamine biosynthetic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0