Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • DTNBP1 Antibodies

Abnova

DTNBP1 Polyclonal Antibody, MaxPab™

View all (16) DTNBP1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite DTNBP1 Polyclonal Antibody, MaxPab™

  • Antibody Testing Data (2)
DTNBP1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
DTNBP1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

DTNBP1 Antibody (H00084062-D01P) in ICC/IF

Immunofluorescence of purified MaxPab antibody to DTNBP1 on HeLa cell. Antibody concentration is 30 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

DTNBP1 Antibody in Immunocytochemistry (ICC/IF)
DTNBP1 Antibody in Western Blot (WB)
DTNBP1 Polyclonal Antibody, MaxPab™

Product Details

H00084062-D01P

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000
-

Immunocytochemistry (ICC/IF)

30 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

DTNBP1 (NP_115498.2, 1 a.a. approximately 351 a.a) full-length human protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Sequence of this protein is as follows: MLETLRERLL SVQQDFTSGL KTLSDKSREA KVKSKPRTVP FLPKYSAGLE LLSRYEDTWA ALHRRAKDCA SAGELVDSEV VMLSAHWEKK KTSLVELQEQ LQQLPALIAD LESMTANLTH LEASFEEVEN NLLHLEDLCG QCELERCKHM QSQQLENYKK NKRKELETFK AELDAEHAQK VLEMEHTQQM KLKERQKFFE EAFQQDMEQY LSTGYLQIAE RREPIGSMSS MEVNVDMLEQ MDLMDISDQE ALDVFLNSGG EENTVLSPAL GPESSTCQNE ITLQVPNPSE LRAKPPSSSS TCTDSATRDI SEGGESPVVQ SDEEEVQVDT ALATSHTDRE ATPDGGEDSD S

Target Information

Dtnbp1 may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. A similar protein in mouse is a component of a protein complex termed biogenesis of lysosome-related organelles complex 1 (BLOC-1), and binds to alpha- and beta-dystrobrevins, which are components of the dystrophin-associated protein complex (DPC). Mutations are associated with Hermansky-Pudlak syndrome type 7. This protein may also be associated with schizophrenia.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: biogenesis of lysosomal organelles complex-1, subunit 8; Biogenesis of lysosome-related organelles complex 1 subunit 8; BLOC-1 subunit 8; DKFZp564K192; DTBP1; Dysbindin; Dysbindin-1; Dystrobrevin-binding protein 1; FLJ30031; Hermansky-Pudlak syndrome 7 protein; HPS7 protein; Hps7-like protein; MGC20210; OTTHUMP00000221462; OTTHUMP00000221485; RP1-147M19.1

View more View less

Gene Aliases: BLOC1S8; DBND; DTNBP1; HPS7; My031; SDY

View more View less

UniProt ID: (Human) Q96EV8

View more View less

Entrez Gene ID: (Human) 84062

View more View less

Function(s)
protein binding
Process(es)
positive regulation of neurotransmitter secretion blood coagulation anterograde axonal transport positive regulation of gene expression regulation of dopamine secretion neuron projection development actin cytoskeleton reorganization melanosome organization regulation of JUN kinase activity anterograde synaptic vesicle transport neuron projection morphogenesis platelet dense granule organization regulation of dopamine receptor signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0