Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • DMP1 Antibodies

Abnova

DMP1 Monoclonal Antibody (1D4)

View all (9) DMP1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite DMP1 Monoclonal Antibody (1D4)

  • Antibody Testing Data (2)
DMP1 Antibody in ELISA (ELISA)
Group 53 Created with Sketch.
DMP1 Antibody in ELISA (ELISA)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

DMP1 Antibody (H00001758-M01) in ELISA

Detection limit for recombinant GST tagged DMP1 is 1 ng/mL as a capture antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

DMP1 Antibody in Immunocytochemistry (ICC/IF)
DMP1 Antibody in ELISA (ELISA)
DMP1 Monoclonal Antibody (1D4)

Product Details

H00001758-M01

Applications
Tested Dilution
Publications

Western Blot (WB)

1-5 µg/mL
-

Immunocytochemistry (ICC/IF)

10 µg/mL
-

ELISA (ELISA)

1 ng/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b, kappa

Class

Monoclonal

Type

Antibody

Clone

1D4

Immunogen

DMP1 (NP_004398.1, 221 a.a. approximately 330 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Sequence of this protein is as follows: ESIRSERGNS RMNSAGMKSK ESGENSEQAN TQDSGGSQLL EHPSRKIFRK SRISEEDDRS ELDDNNTMEE VKSDSTENSN SRDTGLSQPR RDSKGDSQED SKENLSQEES

Target Information

Dentin matrix acidic phosphoprotein is an extracellular matrix protein and a member of the small integrin binding ligand N-linked glycoprotein family. This protein, which is critical for proper mineralization of bone and dentin, is present in diverse cells of bone and tooth tissues. The protein contains a large number of acidic domains, multiple phosphorylation sites, a functional arg-gly-asp cell attachment sequence, and a DNA binding domain. In undifferentiated osteoblasts it is primarily a nuclear protein that regulates the expression of osteoblast-specific genes. During osteoblast maturation the protein becomes phosphorylated and is exported to the extracellular matrix, where it orchestrates mineralized matrix formation. Mutations in the gene are known to cause autosomal recessive hypophosphatemia, a disease that manifests as rickets and osteomalacia. The gene structure is conserved in mammals. Two transcript variants encoding different isoforms have been described for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Dentin matrix acidic phosphoprotein 1; dentin matrix protein 1; DMP 1; DMP-1; MGC130441

View more View less

Gene Aliases: ARHP; ARHR; DMP-1; DMP1

View more View less

UniProt ID: (Human) Q13316

View more View less

Entrez Gene ID: (Human) 1758

View more View less

Function(s)
integrin binding calcium ion binding extracellular matrix binding
Process(es)
ossification positive regulation of cell-substrate adhesion extracellular matrix organization biomineral tissue development
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0