Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • DKK3 Antibodies

Proteintech

DKK3 Polyclonal Antibody, CoraLite® Plus 647

View all (20) DKK3 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite DKK3 Polyclonal Antibody, CoraLite® Plus 647

  • Antibody Testing Data (1)
DKK3 Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
DKK3 Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

DKK3 Antibody (CL647-10365) in Flow

1X10^6 HepG2 cells were intracellularly stained with 0.2 µg CoraLite® Plus 647 Anti-Human DKK3 (CL647-10365) (red), or 0.2 µg isotype control (blue). Cells were fixed with 4% PFA and permeabilized with Flow Cytometry Perm Buffer (PF00011-C). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

DKK3 Antibody in Flow Cytometry (Flow)
DKK3 Polyclonal Antibody, CoraLite® Plus 647

Product Details

CL647-10365

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

0.2 µg/1x10^6 cell suspension
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant protein
View immunogen

Conjugate

CoraLite® Plus 647 CoraLite® Plus 647 CoraLite® Plus 647

Excitation/Emission Max

654/674 nm

Form

Liquid

Purification

Antigen affinity chromatography

Storage buffer

PBS with 50% glycerol, 0.5% BSA

Contains

0.05% ProClin 300

Storage conditions

-20°C, store in dark

Shipping conditions

Wet ice

Product Specific Information

Immunogen sequence: ALSYPQEEAT LNEMFREVEE LMEDTQHKLR SAVEEMEAEE AAAKASSEVN LANLPPSYHN ETNTDTKVGN NTIHVHREIH KITNNQTGQM VFSETVITSV GDEEGRRSHE CIIDEDCGPS MYCQFASFQY TCQPCRGQRM LCTRDSECCG DQLCVWGHCT KMATRGSNGT ICDNQRDCQP GLCCAFQRGL LFPVCTPLPV EGELCHDPAS RLLDLITWEL EPDGALDRCP CASGLLCQPH SHSLVYVCKP TFVGSRDQDG EILLPREVPD EYEVGSFMEE VRQELEDLER SLTEEMALGE PAAAAAALLG GEEI

Target Information

DKK3, like DKK1, DKK2, and DKK4, possesses an N-terminal signal peptide and 2 conserved cysteine-rich domains, which are separated by a linker region and contain 10 cysteine residues each. The second cysteine region has a putative lipid-binding function that may facilitate WNT/DKK interactions at the plasma membrane. The linker region contains 50 to 55 amino acids in DKK1, DKK2, and DKK4, whereas in DKK3 it contains only 12 amino acids. All DKKs have several potential sites for cleavage by furin-type proteases. Northern blot analysis revealed wide expression of the DKK3 transcript, with highest expression in heart, brain, and spinal cord. In situ hybridization reveals highest expression in mouse brain, eye, and heart.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Dickkopf; dickkopf 3; dickkopf 3 homolog; Dickkopf gene 3; dickkopf homolog 3; Dickkopf-3; Dickkopf-related protein 3; Dickkopf3; DKK 3; dkk-3; hDkk 3; hDkk-3; hhDkk3; mDkk-3; regulated in glioma; RIG-like 5-6; RIG-like 7-1; Xenopus

View more View less

Gene Aliases: AW061014; C87148; DKK3; REIC; RIG; UNQ258/PRO295

View more View less

UniProt ID: (Human) Q9UBP4, (Mouse) Q9QUN9

View more View less

Entrez Gene ID: (Human) 27122, (Mouse) 50781

View more View less

Process(es)
anatomical structure morphogenesis Wnt signaling pathway regulation of transforming growth factor beta receptor signaling pathway adrenal gland development negative regulation of aldosterone biosynthetic process negative regulation of transcription, DNA-templated negative regulation of canonical Wnt signaling pathway negative regulation of anti-Mullerian hormone signaling pathway negative regulation of cortisol biosynthetic process multicellular organism development negative regulation of Wnt signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0