Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • DJ-1 Antibodies

Invitrogen

DJ-1 Monoclonal Antibody (4B10)

View all (34) DJ-1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite DJ-1 Monoclonal Antibody (4B10)

  • Antibody Testing Data (6)
DJ-1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
DJ-1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 6

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

DJ-1 Antibody (MA5-49165) in ICC/IF

Immunocytochemistry analysis of DJ-1 in Hela cells. Enzyme antigen retrieval was performed using IHC enzyme antigen retrieval reagent for 15 mins. The cells were blocked with 10% goat serum. Samples were then incubated in DJ-1 Monoclonal antibody (Product # MA5-49165) using a dilution of 5 μg/mL. DyLight®488 Conjugated Goat Anti-Mouse IgG was used as secondary antibody at 1:100 dilution and incubated for 30 minutes at 37°C. The section was counterstained with DAPI. Visualize using a fluorescence microscope and filter sets appropriate for t... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

DJ-1 Antibody in Immunocytochemistry (ICC/IF)
DJ-1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
DJ-1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
DJ-1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
DJ-1 Antibody in Western Blot (WB)
DJ-1 Antibody in Flow Cytometry (Flow)
DJ-1 Monoclonal Antibody (4B10)

Product Details

MA5-49165

Applications
Tested Dilution
Publications

Western Blot (WB)

0.25-0.5 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

2-5 µg/mL
-

Immunocytochemistry (ICC/IF)

5 µg/mL
-

Flow Cytometry (Flow)

1-3 µg/1x10^6 cells
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

4B10

Immunogen

E.coli-derived human PARK7 recombinant protein (Position: A2-D189).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

500 µg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS with 4mg trehalose

Contains

no preservative

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_3074823

Product Specific Information

Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.

Human PARK7 shares 91% amino acid (aa) sequence identity with both mouse and rat PARK7.

Positive Control - WB: human U20S whole cell, human HepG2 whole cell. IHC: human endometrial carcinoma tissue, human esophageal squamous carcinoma tissue, human liver cancer tissue. ICC/IF: Hela cell. Flow: Hela cell.|Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Target Information

The product of this gene belongs to the peptidase C56 family of proteins. It acts as a positive regulator of androgen receptor-dependent transcription. It may also function as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. Defects in this gene are the cause of autosomal recessive early-onset Parkinson disease 7. Two transcript variants encoding the same protein have been identified for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CTA-215D11.1; DJ-1; early onset 7; epididymis secretory sperm binding protein Li 67p; FLJ27376; FLJ34360; FLJ92274; Maillard deglycase; Oncogene DJ1; OTTHUMP00000001350; OTTHUMP00000001351; PARK7 GENE; PARK7/DJ1; Parkinson disease (autosomal recessive, early onset) 7; Parkinson disease protein 7; parkinson protein 7; Parkinsonism-associated deglycase; Protein DJ-1; Protein/nucleic acid deglycase DJ-1

View more View less

Gene Aliases: DJ-1; DJ1; HEL-S-67p; PARK7

View more View less

UniProt ID: (Human) Q99497

View more View less

Entrez Gene ID: (Human) 11315

View more View less

Function(s)
core promoter binding double-stranded DNA binding single-stranded DNA binding transcription coactivator activity mRNA binding receptor binding copper ion binding protein binding transcription factor binding peptidase activity superoxide dismutase copper chaperone activity oxidoreductase activity, acting on peroxide as acceptor glyoxalase III activity enzyme binding kinase binding cytokine binding tyrosine 3-monooxygenase activator activity L-dopa decarboxylase activator activity identical protein binding protein homodimerization activity small protein activating enzyme binding ubiquitin-like protein conjugating enzyme binding mercury ion binding androgen receptor binding peroxiredoxin activity repressing transcription factor binding scaffold protein binding cadherin binding involved in cell-cell adhesion cupric ion binding cuprous ion binding ubiquitin-specific protease binding glyoxalase (glycolic acid-forming) activity
Process(es)
negative regulation of protein phosphorylation negative regulation of protein kinase activity proteolysis protein deglycosylation autophagy inflammatory response mitochondrion organization Ras protein signal transduction single fertilization adult locomotory behavior detoxification of copper ion positive regulation of gene expression negative regulation of gene expression enzyme active site formation via L-cysteine sulfinic acid methylglyoxal catabolic process to D-lactate via S-lactoyl-glutathione lactate biosynthetic process protein repair negative regulation of protein ubiquitination negative regulation of protein binding activation of protein kinase B activity negative regulation of proteasomal ubiquitin-dependent protein catabolic process positive regulation of interleukin-8 production positive regulation of peptidyl-serine phosphorylation negative regulation of protein sumoylation cellular response to oxidative stress cellular response to glyoxal response to drug hydrogen peroxide metabolic process negative regulation of apoptotic process regulation of neuron apoptotic process negative regulation of neuron apoptotic process regulation of TRAIL receptor biosynthetic process positive regulation of transcription from RNA polymerase II promoter glycolate biosynthetic process negative regulation of protein export from nucleus regulation of inflammatory response detoxification of mercury ion protein stabilization positive regulation of sequence-specific DNA binding transcription factor activity negative regulation of ubiquitin-protein transferase activity dopamine uptake involved in synaptic transmission regulation of mitochondrial membrane potential positive regulation of protein kinase B signaling membrane depolarization membrane hyperpolarization negative regulation of cell death regulation of androgen receptor signaling pathway cellular response to hydrogen peroxide positive regulation of protein homodimerization activity cell-cell adhesion cellular oxidant detoxification positive regulation of protein localization to nucleus negative regulation of neuron death positive regulation of superoxide dismutase activity negative regulation of protein acetylation positive regulation of oxidative stress-induced intrinsic apoptotic signaling pathway negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway regulation of fibril organization positive regulation of mitochondrial electron transport, NADH to ubiquinone negative regulation of death-inducing signaling complex assembly negative regulation of protein K48-linked deubiquitination negative regulation of TRAIL-activated apoptotic signaling pathway positive regulation of pyrroline-5-carboxylate reductase activity positive regulation of tyrosine 3-monooxygenase activity positive regulation of dopamine biosynthetic process glyoxal catabolic process positive regulation of L-dopa biosynthetic process positive regulation of L-dopa decarboxylase activity negative regulation of oxidative stress-induced cell death negative regulation of hydrogen peroxide-induced cell death negative regulation of hydrogen peroxide-induced neuron death negative regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway negative regulation of hydrogen peroxide-induced neuron intrinsic apoptotic signaling pathway positive regulation of reactive oxygen species biosynthetic process positive regulation of mitophagy negative regulation of nitrosative stress-induced intrinsic apoptotic signaling pathway negative regulation of ubiquitin-specific protease activity positive regulation of oxidative phosphorylation uncoupler activity positive regulation of transcription regulatory region DNA binding positive regulation of androgen receptor activity negative regulation of extrinsic apoptotic signaling pathway negative regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0