Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • DCPS Antibodies

OriGene

DCPS Monoclonal Antibody (OTI4H8), TrueMAB™

View all (16) DCPS antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite DCPS Monoclonal Antibody (OTI4H8), TrueMAB™

  • Antibody Testing Data (11)
DCPS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
DCPS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 11

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

DCPS Antibody (CF505626) in IHC (P)

Immunohistochemical staining of paraffin-embedded human lymphoma tissue using anti-DCPS mouse monoclonal antibody. (Heat-induced epitope retrieval by 10mM citric buffer, pH6.0, 120°C for 3min, TA505626) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

DCPS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
DCPS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
DCPS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
DCPS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
DCPS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
DCPS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
DCPS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
DCPS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
DCPS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
DCPS Antibody in Western Blot (WB)
DCPS Antibody in Western Blot (WB)
DCPS Monoclonal Antibody (OTI4H8), TrueMAB™

Product Details

CF505626

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:150
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI4H8

Immunogen

Full length human recombinant protein of human DCPS produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

Necessary for the complete degradation of mRNAs, both in normal mRNA turnover and in nonsense-mediated mRNA decay. Removes the 7-methyl guanine cap structure from mRNA fragments shorter than 10 nucleotides that are produced by 3'->5' exosome-mediated mRNA decay. Releases m7GMP. Can also degrade m7GDP to m7GMP. Has no activity towards mRNA molecules longer than 25 nucleotides.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 5'-(N(7)-methyl 5'-triphosphoguanosine)-[mRNA] diphosphatase; DCS-1; Decapping scavenger enzyme; heat shock-like protein 1; HINT-5; Hint-related 7meGMP-directed hydrolase; Histidine triad nucleotide-binding protein 5; Histidine triad protein member 5; homolog of C. elegans 7meGMP-directed hydrolase dcs-1; m7GpppX diphosphatase; mRNA decapping enzyme; Scavenger mRNA-decapping enzyme DcpS

View more View less

Gene Aliases: ARS; DCPS; DCS1; HINT-5; HINT5; HSL1; HSPC015

View more View less

UniProt ID: (Human) Q96C86

View more View less

Entrez Gene ID: (Human) 28960

View more View less

Function(s)
RNA 7-methylguanosine cap binding exoribonuclease activity protein binding m7G(5')pppN diphosphatase activity
Process(es)
nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay deadenylation-dependent decapping of nuclear-transcribed mRNA cellular response to menadione negative regulation of programmed cell death exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay mRNA cis splicing, via spliceosome RNA phosphodiester bond hydrolysis, exonucleolytic
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0