Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • D-dimer Antibodies

OriGene

D-Dimer Monoclonal Antibody (OTI7F12), TrueMAB™

View all (63) D-dimer antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite D-Dimer Monoclonal Antibody (OTI7F12), TrueMAB™

D-Dimer Monoclonal Antibody (OTI7F12), TrueMAB™

Product Details

CF813865

Applications
Tested Dilution
Publications

ELISA (ELISA)

1:5,000-1:10,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI7F12

Immunogen

Native Human D-Dimer protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

See Label

Purification

Protein A/G

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

Fibrinogen is the main protein of blood coagulation system. It consists of two identical subunits that contain three polypeptide chains: alpha, beta and gamma. The process of blood coagulation results in the activation of fibrinogen into fibrin by thrombin and fibrin polymerization. Fibrin clot is then digested by plasmin, and fibrin degradation products of different molecular weights are released into the bloodstream.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Fibrinogen alpha chain; fibrinogen, A alpha polypeptide; MGC119422; MGC119423; MGC119425

View more View less

Gene Aliases: FGA; Fib2

View more View less

UniProt ID: (Human) P02671

View more View less

Entrez Gene ID: (Human) 2243

View more View less

Function(s)
receptor binding structural molecule activity protein binding protein binding, bridging metal ion binding cell adhesion molecule binding
Process(es)
adaptive immune response platelet degranulation protein complex assembly acute-phase response cell-matrix adhesion signal transduction blood coagulation extracellular matrix organization plasminogen activation response to estradiol response to genistein positive regulation of heterotypic cell-cell adhesion fibrinolysis induction of bacterial agglutination response to morphine cellular protein complex assembly cellular protein metabolic process innate immune response positive regulation of vasoconstriction positive regulation of exocytosis response to cycloheximide positive regulation of protein secretion protein polymerization response to calcium ion positive regulation of ERK1 and ERK2 cascade platelet aggregation cellular response to interleukin-6 cellular response to organic cyclic compound blood coagulation, common pathway blood coagulation, fibrin clot formation positive regulation of peptide hormone secretion liver regeneration positive regulation of substrate adhesion-dependent cell spreading negative regulation of extrinsic apoptotic signaling pathway via death domain receptors cellular response to granulocyte colony-stimulating factor negative regulation of endothelial cell apoptotic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0