Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Cytokeratin 5/8 Antibodies

Progen

Cytokeratin 5/8 Antibody Cocktail, Biotin

View all (15) Cytokeratin 5/8 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Cytokeratin 5/8 Antibody Cocktail, Biotin

Product Details

61531PROGEN

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1:10-1:20
-

Immunohistochemistry (Frozen) (IHC (F))

1:10-1:20
-

Immunocytochemistry (ICC/IF)

Assay-dependent
-

ELISA (ELISA)

Assay-dependent
-
Product Specifications

Species Reactivity

Bovine, Human, Mouse

Host/Isotype

Mouse / IgG1

Class

Cocktail

Type

Antibody

Clone

Ks 5+8.22/C22

Immunogen

Human Keratin K8, Purified From Sds Page Gel

Conjugate

Biotin Biotin Biotin

Form

Liquid

Purification

Affinity chromatography

Contains

0.09% sodium azide

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

61531 is a mixture of monoclonal antibodies that target Cytokeratin 5/8.

C 22 represents an excellent marker for distinguishing carcinomas from all non-epithelial tumors. The antibody specifically reacts with keratins K5 and K8 present in nearly all epithelia. Polypeptide reacting: MW 52 500, MW 58 000 keratins (type II keratins K5 and K8; formerly also designated cytokeratins 5 and 8) of human epithelial cells. This antibody was used in a Direct ELISA.

Target Information

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between ~40 kDa and ~68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 58 kDa cytokeratin; CK-5; CK-8; cytokeratin 5; cytokeratin 8; Cytokeratin endo A; Cytokeratin-5; Cytokeratin-8; Cytokeratin-A; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; K5; K8; KERA; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 8, type II; keratin complex 2, basic, gene 5; keratin complex 2, basic, gene 8; Keratin, type II cytoskeletal 5; keratin, type II cytoskeletal 60 kDa, component III; Keratin, type II cytoskeletal 8; Keratin-5; Keratin-8; krt5; tuftelin interacting protein 8; Type-II keratin Kb5; Type-II keratin Kb8

View more View less

Gene Aliases: 3300001P10Rik; AA960620; AL022697; AU019895; AW146334; BOS_5040; BOS_5041; CARD2; CK-8; CK5; CK8; CYK8; DDD; DDD1; EBS2; EndoA; K2C8; K5; K8; KO; Krt-2.8; Krt2-5; KRT2-8; KRT5; KRT5A; KRT8; Tfip8

View more View less

UniProt ID: (Bovine) Q5XQN5, (Human) P13647, (Bovine) P05786, (Human) P05787, (Mouse) Q922U2, (Mouse) P11679

View more View less

Entrez Gene ID: (Bovine) 281268, (Human) 3852, (Bovine) 281269, (Human) 3856, (Mouse) 110308, (Mouse) 16691

View more View less

Function(s)
structural molecule activity structural constituent of cytoskeleton protein binding scaffold protein binding protein complex binding
Process(es)
epithelial cell differentiation negative regulation of epithelial cell proliferation cell morphogenesis involved in differentiation multicellular organism development tumor necrosis factor-mediated signaling pathway sarcomere organization response to hydrostatic pressure response to other organism cell differentiation involved in embryonic placenta development extrinsic apoptotic signaling pathway hepatocyte apoptotic process viral process cytoskeleton organization epidermis development hemidesmosome assembly
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0