Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Cytokeratin 18 Antibodies

Invitrogen

Cytokeratin 18 Recombinant Rabbit Monoclonal Antibody (7B2N3)

View all (157) Cytokeratin 18 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Cytokeratin 18 Recombinant Rabbit Monoclonal Antibody (7B2N3)

  • Antibody Testing Data (7)
Cytokeratin 18 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Cytokeratin 18 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 7

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Cytokeratin 18 Antibody (MA5-37993) in IHC (P)

Immunohistochemistry analysis of Cytokeratin 18 in paraffin-embedded human liver cancer. Samples were incubated with Cytokeratin 18 Monoclonal antibody (Product # MA5-37993) using a dilution of 1:100 (40x lens). Perform microwave antigen retrieval with 10 mM Tris/EDTA buffer pH 9.0 before commencing with IHC staining protocol. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Cytokeratin 18 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Cytokeratin 18 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Cytokeratin 18 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Cytokeratin 18 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Cytokeratin 18 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Cytokeratin 18 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Cytokeratin 18 Antibody in Western Blot (WB)
Cytokeratin 18 Recombinant Rabbit Monoclonal Antibody (7B2N3)

Product Details

MA5-37993

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000-1:6,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:500-1:2,000
-

ELISA (ELISA)

1 µg/mL
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

7B2N3

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 250-430 of human Cytokeratin 18 (KRT18) (P05783)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.6 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol, 0.05% BSA

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2897911

Product Specific Information

Positive Samples: HeLa, HepG2, A-431

Immunogen sequence: ADIRAQYDEL ARKNREELDK YWSQQIEEST TVVTTQSAEV GAAETTLTEL RRTVQSLEID LDSMRNLKAS LENSLREVEA RYALQMEQLN GILLHLESEL AQTRAEGQRQ AQEYEALLNI KVKLEAEIAT YRRLLEDGED FNLGDALDSS NSMQTIQKTT TRRIVDGKVV SETNDTKVLR H

Target Information

Cytokeratin 18 (Keratin 18, KRT18, type I cytoskeletal 18) belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Further, cytokeratin 18 is found primarily in non-squamous epithelia and is present in a majority of adenocarcinomas and ductal carcinomas, but not in squamous cell carcinomas. Hepatocellular carcinomas express only cytokeratins 8 and 18. Mutations in the cytokeratin 18 gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for the cytokeratin 18 gene. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Cell proliferation-inducing gene 46 protein; CK 18; CK-18; cytokeratin 18; Cytokeratin endo B; Cytokeratin-18; Cytokeratin18; Endo B; K 18; K18; keratin 18, type I; keratin complex 1, acidic, gene 18; Keratin D; Keratin, type I cytoskeletal 18; Keratin-18; Keratin18; KRT 18; krt18 {ECO:0000250|UniProtKB:P05783}; type I cytoskeletal 18

View more View less

Gene Aliases: CK-18; CK18; CYK18; K18; Kerd; Krt1-18; KRT18; PIG46

View more View less

UniProt ID: (Human) P05783, (Rat) Q5BJY9, (Mouse) P05784

View more View less

Entrez Gene ID: (Human) 3875, (Rat) 294853, (Mouse) 16668

View more View less

Function(s)
structural molecule activity protein binding poly(A) RNA binding scaffold protein binding cadherin binding involved in cell-cell adhesion molecular_function
Process(es)
cell cycle anatomical structure morphogenesis viral process tumor necrosis factor-mediated signaling pathway Golgi to plasma membrane CFTR protein transport negative regulation of apoptotic process intermediate filament cytoskeleton organization extrinsic apoptotic signaling pathway hepatocyte apoptotic process cell-cell adhesion biological_process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0