Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Connexin 32 Antibodies

Invitrogen

Connexin 32 Recombinant Rabbit Monoclonal Antibody (23GB5895)

View all (22) Connexin 32 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Connexin 32 Recombinant Rabbit Monoclonal Antibody (23GB5895)

  • Antibody Testing Data (2)
Connexin 32 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
Connexin 32 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Connexin 32 Antibody (MA5-52942) in WB

Western Blot analysis of Connexin 32. Total cell lysates (30 μg) from various cell lines were loaded and separated by SDS-PAGE. The blot was incubated with Connexin 32 recombinant monoclonal antibody (Product # MA5-52942) with a dilution of 1:5,000, followed by HRP-conjugated goat anti-rabbit secondary antibody with a dilution of 1:20,000. Image was developed using FeQTM ECL Substrate Kit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Connexin 32 Antibody in Western Blot (WB)
Connexin 32 Antibody in Flow Cytometry (Flow)
Connexin 32 Recombinant Rabbit Monoclonal Antibody (23GB5895)

Product Details

MA5-52942

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000-1:5,000
-

Flow Cytometry (Flow)

1:2,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293F

Class

Recombinant Monoclonal

Type

Antibody

Clone

23GB5895

Immunogen

A synthesized peptide derived from human GJB1 (50-84AA).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_3249417

Target Information

This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: connexin 32; Connexin-32; Cx32; GAP junction 28 kDa liver protein; Gap junction beta-1 protein; gap junction protein, beta 1, 32kDa

View more View less

Gene Aliases: CMTX; CMTX1; CX32; GJB1

View more View less

UniProt ID: (Human) P08034

View more View less

Entrez Gene ID: (Human) 2705

View more View less

Function(s)
gap junction channel activity protein homodimerization activity
Process(es)
transport cell-cell signaling nervous system development purine ribonucleotide transport gap junction assembly protein oligomerization transmembrane transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0