Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Complement C4d Antibodies

NeoBiotechnologies

Complement 4d (C4d) (Acute Humoral Rejection Marker) Monoclonal Antibody (SPM545)

View all (20) Complement C4d antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Complement 4d (C4d) (Acute Humoral Rejection Marker) Monoclonal Antibody (SPM545)

  • Antibody Testing Data (1)
Complement 4d (C4d) (Acute Humoral Rejection Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Complement 4d (C4d) (Acute Humoral Rejection Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Complement 4d (C4d) (Acute Humoral Rejection Marker) Antibody (720-MSM1X-P1ABX) in IHC (P)

Formalin-fixed, paraffin-embedded human Kidney Transplant stained with Complement 4d Monoclonal Antibody (SPM545). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Complement 4d (C4d) (Acute Humoral Rejection Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Complement 4d (C4d) (Acute Humoral Rejection Marker) Monoclonal Antibody (SPM545)

Product Details

720-MSM1X-P1ABX

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1:200-1:400
-

Immunocytochemistry (ICC/IF)

1:50-1:100
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

SPM545

Immunogen

Recombinant human Complement 4d protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

C4 is a component of the classical complement pathway and is split by activated C1s to C4a and C4b. Together C2a and C4b forms the classical C3 convertase. The molecular mass of C4 is 204 kDa and it consists of 3 subunits: a (97 kDa), b (75 kDa) and g (33 kDa).

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: acidic C4; Acidic complement C4; Basic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3; c4 complement; C4A anaphylatoxin; C4D; Chido form of C4; Complement C4-A; Complement C4-B; complement C4B1a; DADB-112B14.11; FLJ60561; MGC164979; MHC class III region complement; Rodgers form of C4

View more View less

Gene Aliases: C4; C4A; C4A2; C4A3; C4A4; C4A6; C4AD; C4B; C4B1; C4B12; C4B2; C4B3; C4B5; C4B_2; C4BD; C4F; C4S; CH; CO4; CPAMD2; CPAMD3; RG

View more View less

UniProt ID: (Human) P0C0L4, (Human) P0C0L5

View more View less

Entrez Gene ID: (Human) 720, (Human) 721, (Human) 100293534

View more View less

Function(s)
complement component C1q binding serine-type endopeptidase activity endopeptidase inhibitor activity complement binding carbohydrate binding
Process(es)
proteolysis inflammatory response complement activation complement activation, classical pathway negative regulation of endopeptidase activity regulation of complement activation innate immune response positive regulation of apoptotic cell clearance opsonization detection of molecule of bacterial origin
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0