Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Collagen I Antibodies

Proteintech

Collagen Type I Monoclonal Antibody (1E9A7), CoraLite® Plus 488

View all (24) Collagen I antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Collagen Type I Monoclonal Antibody (1E9A7), CoraLite® Plus 488

  • Antibody Testing Data (2)
Collagen Type I Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Collagen Type I Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Collagen Type I Antibody (CL48867288100UL) in IHC (P)

Immunofluorescent analysis of (4% PFA) fixed human colon cancer tissue using CoraLite 488 Collagen Type I antibody (Product # CL48867288100UL), Clone: 1E9A7 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Collagen Type I Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Collagen Type I Antibody in Immunohistochemistry (Paraffin) (IHC (P))

Product Details

CL48867288100UL

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1:200
-

Immunocytochemistry (ICC/IF)

1:50-1:500
-
Product Specifications

Species Reactivity

Human, Pig

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

1E9A7

Conjugate

CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
  • Unconjugated
  • CoraLite 594
  • Request custom conjugation

Excitation/Emission Max

493/522 nm

Form

Liquid

Concentration

1 mg/mL

Purification

Protein G

Storage buffer

PBS, pH 7.3, with 0.5% BSA, 50% glycerol

Contains

0.05% ProClin 300

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles, store in dark

Shipping conditions

Wet ice

Target Information

Collagen I is a Type I collagen with a triple helix structure comprised of two alpha-1 chains and one alpha-2 chain. Collagen I is a member of group I collagen (fibril-forming collagen) found in most connective tissues, and is abundant in bone, cornea, dermis and tendon. Mutations in the COL1A2 gene encoding the alpha-2 chain are associated with osteogenesis imperfecta types I-IV, Ehlers-Danlos syndrome type VIIB, recessive Ehlers-Danlos syndrome Classical type, idiopathic osteoporosis, and atypical Marfan syndrome. Symptoms associated with mutations in the COL1A2 gene which codes for the alpha-2 chain, however, tend to be less severe than mutations in the COL1A1 gene, reflecting the different role of alpha-2 chains in matrix integrity. Reciprocal translocations between chromosomes 17 and 22, where Collagen Type 1 genes and the gene for platelet derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: alpha 2 type I procollagen; alpha 2(I) procollagen; alpha 2(I)-collagen; Alpha-1 type I collagen; Alpha-2 type I collagen; alpha1(I) procollagen; COL1; collagen alpha 1 chain type I; Collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; Collagen alpha-2(I) chain; collagen I, alpha-2 polypeptide; collagen of skin, tendon and bone, alpha-1 chain; collagen of skin, tendon and bone, alpha-2 chain; collagen, type I, alpha 1; collagen, type I, alpha 2; pro-alpha-1 collagen type 1; Type I Collagen; type I proalpha 1; type I procollagen; type I procollagen alpha 1 chain

View more View less

Gene Aliases: COL1A1; COL1A2; EDSC; OI1; OI2; OI3; OI4

View more View less

UniProt ID: (Human) P02452, (Human) P08123

View more View less

Entrez Gene ID: (Human) 1277, (Human) 1278, (Pig) 100626716

View more View less

Function(s)
extracellular matrix structural constituent protein binding identical protein binding metal ion binding platelet-derived growth factor binding protein binding, bridging SMAD binding
Process(es)
skeletal system development blood vessel development transforming growth factor beta receptor signaling pathway Rho protein signal transduction blood coagulation regulation of blood pressure platelet activation extracellular matrix organization collagen fibril organization collagen catabolic process odontogenesis skin morphogenesis regulation of immune response leukocyte migration protein heterotrimerization cellular response to amino acid stimulus osteoblast differentiation intramembranous ossification endochondral ossification visual perception sensory perception of sound positive regulation of epithelial to mesenchymal transition negative regulation of cell-substrate adhesion protein transport positive regulation of cell migration response to corticosteroid response to estradiol collagen biosynthetic process protein localization to nucleus tooth mineralization response to drug response to hydrogen peroxide response to peptide hormone cellular response to fibroblast growth factor stimulus tooth eruption positive regulation of transcription, DNA-templated embryonic skeletal system development response to cAMP response to hyperoxia face morphogenesis bone trabecula formation cartilage development involved in endochondral bone morphogenesis cellular response to mechanical stimulus cellular response to retinoic acid cellular response to vitamin E cellular response to tumor necrosis factor cellular response to epidermal growth factor stimulus cellular response to transforming growth factor beta stimulus positive regulation of canonical Wnt signaling pathway cellular response to fluoride
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0