Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Cathepsin D Antibodies

Invitrogen

Cathepsin D Polyclonal Antibody

View all (56) Cathepsin D antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Cathepsin D Polyclonal Antibody

  • Antibody Testing Data (4)
Cathepsin D Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Cathepsin D Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Cathepsin D Antibody (PA5-89138) in IHC (P)

Immunohistochemistry analysis of Cathepsin D in paraffin-embedded human tonsil. Samples were incubated with Cathepsin D Polyclonal antibody (Product # PA5-89138) using a dilution of 1:20 (40x lens). Perform high pressure antigen retrieval with 10 mM citrate buffer pH 6.0 before commencing with IHC staining protocol. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Cathepsin D Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Cathepsin D Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Cathepsin D Antibody in Western Blot (WB)
Cathepsin D Antibody in Western Blot (WB)
Cathepsin D Polyclonal Antibody

Product Details

PA5-89138

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-

ELISA (ELISA)

1 µg/mL
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 21-249 of human Cathepsin D (NP_0019001)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1.32 mg/mL

Purification

Affinity Chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.09% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

RRID

AB_2805380

Product Specific Information

Immunogen sequence: LVRIPLHKFT SIRRTMSEVG GSVEDLIAKG PVSKYSQAVP AVTEGPIPEV LKNYMDAQYY GEIGIGTPPQ CFTVVFDTGS SNLWVPSIHC KLLDIACWIH HKYNSDKSST YVKNGTSFDI HYGSGSLSGY LSQDTVSVPC QSASSASALG GVKVERQVFG EATKQPGITF IAAKFDGILG MAYPRISVNN VLPVFDNLMQ QKLVDQNIFS FYLSRDPDAQ PGGELMLGG

Positive Samples: Mouse kidney, Mouse heart; Cellular Location: Lysosome, Melanosome, extracellular space

Target Information

Cathepsin D has been reported to be involved in malignant tumor progression and with prognosis of various human cancers.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Cathepsin D; CathepsinD; Ceroid lipofuscinosis neuronal 10; ceroid-lipofuscinosis, neuronal 10; epididymis secretory sperm binding protein Li 130P; lysosomal aspartyl peptidase; lysosomal aspartyl protease; MGC2311; OTTHUMP00000196039; OTTHUMP00000198692

View more View less

Gene Aliases: CatD; CD; CLN10; CPSD; CTSD; HEL-S-130P

View more View less

UniProt ID: (Human) P07339, (Mouse) P18242

View more View less

Entrez Gene ID: (Human) 1509, (Mouse) 13033, (Rat) 171293

View more View less

Function(s)
aspartic-type endopeptidase activity cysteine-type endopeptidase activity serine-type endopeptidase activity protein binding endopeptidase activity peptidase activity hydrolase activity peptide binding
Process(es)
proteolysis autophagy antigen processing and presentation of exogenous peptide antigen via MHC class II protein catabolic process collagen catabolic process autophagosome assembly epithelial tube branching involved in lung morphogenesis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0