Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Cathelicidin-5 Antibodies

Fabgennix

Cathelicidin-5 Monoclonal Antibody (M48), FITC

View all (6) Cathelicidin-5 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Cathelicidin-5 Monoclonal Antibody (M48), FITC

Cathelicidin-5 Monoclonal Antibody (M48), FITC

Product Details

BMAP28-101MAB-FITC

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500
-

ELISA (ELISA)

1:10,000
-

Immunoprecipitation (IP)

1:200
-
Product Specifications

Species Reactivity

Bovine

Host/Isotype

Mouse / IgG

Class

Monoclonal

Type

Antibody

Clone

M48

Immunogen

Synthetic peptide corresponding to amino acids within region 125-159 on bovine Cathelicidin-5 protein.
View immunogen

Conjugate

FITC FITC FITC

Excitation/Emission Max

498/517 nm View spectra spectra

Form

Liquid

Concentration

0.5-1.5 µg/mL

Purification

Protein G

Storage buffer

proprietary buffer with 30% glycerol

Contains

0.01% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

Exerts a potent antimicrobial activity against Gram-negative and Gram-positive bacteria, including methicillin-resistant Staphylococcus aureus, and fungi.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Antibacterial peptide BMAP-28; antimicrobial peptide BMAP-28; BMAP28; Cathelicidin-5; CATHL5; Myeloid antibacterial peptide 28

View more View less

Gene Aliases: BMAP28; BOS_20928; CATHL5

View more View less

UniProt ID: (Bovine) P54229

View more View less

Entrez Gene ID: (Bovine) 282167

View more View less

Process(es)
defense response to bacterium
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0