Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Cystatin C Antibodies

OriGene

CST3 Monoclonal Antibody (OTI2C11), Biotin, TrueMAB™

View all (81) Cystatin C antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CST3 Monoclonal Antibody (OTI2C11), Biotin, TrueMAB™

  • Antibody Testing Data (1)
CST3 Antibody in Luminex (LUM)
Group 53 Created with Sketch.
CST3 Antibody in Luminex (LUM)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CST3 Antibody (TA700252) in LUM

CST3 Luminex Elisa with anti-Tag 1B12 Capture (TA600050) and 2C11 Detection (TA700252) Antibodies. Substrate used: Recombinant Human CST3 (TP310730) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CST3 Antibody in Luminex (LUM)
CST3 Monoclonal Antibody (OTI2C11), Biotin, TrueMAB™

Product Details

TA700252

Applications
Tested Dilution
Publications

Luminex (LUM)

Assay-Dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI2C11

Immunogen

Full length human recombinant protein of human CST3 produced in HEK293T cell.
View immunogen

Conjugate

Biotin Biotin Biotin
  • Unconjugated
  • Request custom conjugation

Form

Liquid

Concentration

0.5 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 10mg/mL BSA, 50% glycerol

Contains

0.05% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

Cystatin C, a member of the type 2 cystatin superfamily, is a potent inhibitor of cysteine proteinases, both endogenous (lysosomal cathepsins), and pathogenic. Cystatin C is a 13 kDa secreted protein which is found in high concentrations in biological fluids and is thought to be expressed by all human nucleated cells. Changes in Cystatin C have been documented in a number of cardiovascular diseases, including atherosclerosis, amyloid angiopathy and myocardial infarction. Its low molecular weight also means that Cystatin C can be used as a marker of renal failure.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: AD8; bA218C14.4 (cystatin C); cystatin 3; Cystatin-3; Cystatin-C; Cystatin3; CystatinC; epididymis secretory protein Li 2; Gamma-trace; MGC117328; Neuroendocrine basic polypeptide; OTTHUMP00000030440; OTTHUMP00000164181; OTTHUMP00000164182; OTTMUSP00000016736; Post-gamma-globulin; RP23-250M11.9

View more View less

Gene Aliases: ARMD11; CST3; HEL-S-2

View more View less

UniProt ID: (Human) P01034

View more View less

Entrez Gene ID: (Human) 1471

View more View less

Function(s)
beta-amyloid binding protease binding endopeptidase inhibitor activity cysteine-type endopeptidase inhibitor activity protein binding identical protein binding
Process(es)
defense response negative regulation of peptidase activity negative regulation of collagen catabolic process negative regulation of extracellular matrix disassembly negative regulation of endopeptidase activity regulation of tissue remodeling extracellular fibril organization cellular protein metabolic process negative regulation of proteolysis negative regulation of elastin catabolic process negative regulation of blood vessel remodeling
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0