Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CRYAB Antibodies

OriGene

CRYAB Monoclonal Antibody (OTI3C2), TrueMAB™

View all (84) CRYAB antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CRYAB Monoclonal Antibody (OTI3C2), TrueMAB™

  • Antibody Testing Data (10)
CRYAB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
CRYAB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 10

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CRYAB Antibody (CF500573) in IHC (P)

Immunohistochemical staining of paraffin-embedded human lymphoma tissue using anti-CRYAB mouse monoclonal antibody. (Heat-induced epitope retrieval by 10mM citric buffer, pH6.0, 100°C for 10min, TA500573) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CRYAB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CRYAB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CRYAB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CRYAB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CRYAB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CRYAB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CRYAB Antibody in Western Blot (WB)
CRYAB Antibody in Western Blot (WB)
CRYAB Antibody in Flow Cytometry (Flow)
CRYAB Antibody in Immunoprecipitation (IP)
CRYAB Monoclonal Antibody (OTI3C2), TrueMAB™

Product Details

CF500573

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50
-

Flow Cytometry (Flow)

1:100
-

Immunoprecipitation (IP)

2 µg/500 µL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI3C2

Immunogen

Full length human recombinant protein of human CRYAB produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

Lens proteins consist almost entirely of crystallins (about 95%). Crystallins are also found vertebrate skeletal muscle tissue. In the lens, their structural function is to assist in maintaining the proper refractive index of the lens. The mammalian lens contains 3 major classes of crystallins: alpha, beta, and gamma. Alpha-crystallin is the largest of the crystallins and is composed of 2 primary gene products-alpha-A and alpha-B. There are at least 5 different proteins comprising the beta-crystallins. The gamma-crystallins are monomeric, but there are at least 5 gamma crystallins identified in bovine and rat lens. Alpha-Crystallin comprises 40% of total lens protein composition. In addition to maintaining proper refractive index, it also functions in a chaperone like manner by preventing the formation of aggregates possibly leading to cataract formation. It is believed that the phosphorylated states of the alpha-crystallin occur in response to cellular stress and may serve a structural control function and play a role in protein maintenance. Alpha-B crystallin has been linked to Alexander's disease where it accumulates in brain cells of those afflicted.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Alpha(B)-crystallin; Alpha-crystallin B chain; CryAB antibody; crystallin, alpha B; epididymis secretory protein Li 101; Heat shock protein beta-5; Heat shock protein family B member 5; heat-shock 20 kD like-protein; HspB5; Renal carcinoma antigen NY-REN-27; Rosenthal fiber component

View more View less

Gene Aliases: CMD1II; CRYA2; CRYAB; CTPP2; CTRCT16; HEL-S-101; HSPB5; MFM2

View more View less

UniProt ID: (Human) P02511

View more View less

Entrez Gene ID: (Human) 1410

View more View less

Function(s)
structural constituent of eye lens protein binding microtubule binding identical protein binding protein homodimerization activity metal ion binding unfolded protein binding
Process(es)
response to hypoxia lens development in camera-type eye protein folding muscle contraction tubulin complex assembly muscle organ development aging negative regulation of gene expression regulation of cell death negative regulation of cell growth microtubule polymerization or depolymerization response to estradiol negative regulation of intracellular transport response to hydrogen peroxide negative regulation of apoptotic process negative regulation of cysteine-type endopeptidase activity involved in apoptotic process protein homooligomerization stress-activated MAPK cascade apoptotic process involved in morphogenesis cellular response to gamma radiation regulation of cellular response to heat negative regulation of reactive oxygen species metabolic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0