Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CIAPIN1 Antibodies

Abnova

CIAPIN1 Polyclonal Antibody, MaxPab™

View all (32) CIAPIN1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CIAPIN1 Polyclonal Antibody, MaxPab™

  • Antibody Testing Data (2)
CIAPIN1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
CIAPIN1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CIAPIN1 Antibody (H00057019-D01P) in WB

Western Blot analysis of CIAPIN1 expression in transfected 293T cell line (H00057019-T02) by CIAPIN1 MaxPab polyclonal antibody.

Lane 1: CIAPIN1 transfected lysate(33.60 KDa).
Lane 2: Non-transfected lysate.
{{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CIAPIN1 Antibody in Western Blot (WB)
CIAPIN1 Antibody in Western Blot (WB)
CIAPIN1 Polyclonal Antibody, MaxPab™

Product Details

H00057019-D01P

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

CIAPIN1 (NP_064709.2, 1 a.a. approximately 312 a.a) full-length human protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Sequence of this protein is as follows: MADFGISAGQ FVAVVWDKSS PVEALKGLVD KLQALTGNEG RVSVENIKQL LQSAHKESSF DIILSGLVPG STTLHSAEIL AEIARILRPG GCLFLKEPVE TAVDNNSKVK TASKLCSALT LSGLVEVKEL QREPLTPEEV QSVREHLGHE SDNLLFVQIT GKKPNFEVGS SRQLKLSITK KSSPSVKPAV DPAAAKLWTL SANDMEDDSM DLIDSDELLD PEDLKKPDPA SLRAASCGEG KKRKACKNCT CGLAEELEKE KSREQMSSQP KSACGNCYLG DAFRCASCPY LGMPAFKPGE KVLLSDSNLH DA

Target Information

CIAPIN1 is a cytokine-induced inhibitor of apoptosis with no relation to apoptosis regulatory molecules of the BCL2 or CASP families. Expression of CIAPIN1 is dependent on growth factor stimulation.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Anamorsin; Cytokine-induced apoptosis inhibitor 1; Fe-S cluster assembly protein DRE2 homolog; predicted protein of HQ0915

View more View less

Gene Aliases: 2810413N20Rik; AA617265; Anamorsin; AU021794; CIAPIN1; CUA001; DRE2; PRO0915

View more View less

UniProt ID: (Human) Q6FI81, (Mouse) Q8WTY4

View more View less

Entrez Gene ID: (Human) 57019, (Mouse) 109006

View more View less

Function(s)
protein binding methyltransferase activity electron carrier activity metal ion binding 2 iron, 2 sulfur cluster binding iron-sulfur cluster binding
Process(es)
apoptotic process iron-sulfur cluster assembly hemopoiesis methylation negative regulation of apoptotic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0