Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD99 Antibodies

Abnova

CD99 Monoclonal Antibody (3A10)

View all (133) CD99 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD99 Monoclonal Antibody (3A10)

  • Antibody Testing Data (1)
CD99 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
CD99 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD99 Antibody (H00004267-M01) in WB

CD99 monoclonal antibody (M01), clone 3A10. Western Blot analysis of CD99 expression in K-562. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD99 Antibody in Western Blot (WB)
CD99 Monoclonal Antibody (3A10)

Product Details

H00004267-M01

Applications
Tested Dilution
Publications

Western Blot (WB)

1-5 µg/mL
-

ELISA (ELISA)

10 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2a, kappa

Class

Monoclonal

Type

Antibody

Clone

3A10

Immunogen

CD99 (AAH03147, 23 a.a. approximately 122 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

Product Specific Information

Product may be used with Western Blot (Cell lysate).

Immunogen sequence: DGGFDLSDAL PDNENKKPTA IPKKPSAGDD FDLGDAVVDG ENDDPRPPNP PKPMPNPNPN HPSSSGSFSD ADLADGVSGG EGKGGSDGGG SHRKEGEEAD

Target Information

CD99 (E2, MIC2) is a transmembrane glycoprotein that is involved in regulation of T cell addhesive properties and programmed cell death distinct from typical apoptosis course. CD99 roles are specific to certain stages of T cell differentiation such as corticothymocytes. CD99R isoform expression is restricted in the haematopoietic system to T, NK and myeloid cells.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 12E7; antigen identified by monoclonal 12E7, Y homolog; antigen identified by monoclonal antibodies 12E7, F21 and O13; CD99; CD99 antigen; cell surface antigen 12E7; cell surface antigen HBA-71; cell surface antigen O13; E2 antigen; MIC2 (monoclonal antibody 12E7); Protein MIC2; surface antigen MIC2; T-cell surface glycoprotein E2

View more View less

Gene Aliases: CD99; HBA71; MIC2; MIC2X; MIC2Y; MSK5X

View more View less

UniProt ID: (Human) P14209

View more View less

Entrez Gene ID: (Human) 4267

View more View less

Process(es)
cell adhesion
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0