Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD74 Antibodies

NeoBiotechnologies

CD74 (B-Cell Marker) Monoclonal Antibody (LN-2)

View all (91) CD74 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD74 (B-Cell Marker) Monoclonal Antibody (LN-2)

  • Antibody Testing Data (5)
CD74 (B-Cell Marker) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
CD74 (B-Cell Marker) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD74 (B-Cell Marker) Antibody (972-MSM1-P0) in ICC/IF

Immunofluorescence analysis of Raji cells labeling CD74 with CD74 Mouse Monoclonal Antibody (LN-2) followed by Goat anti-Mouse IgG-CF488 (Green). The nuclear counterstain is RedDot (Red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD74 (B-Cell Marker) Antibody in Immunocytochemistry (ICC/IF)
CD74 (B-Cell Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CD74 (B-Cell Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CD74 (B-Cell Marker) Antibody in Western Blot (WB)
CD74 (B-Cell Marker) Antibody in Flow Cytometry (Flow)
CD74 (B-Cell Marker) Monoclonal Antibody (LN-2)

Product Details

972-MSM1-P0

Applications
Tested Dilution
Publications

Western Blot (WB)

2-4 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1-2 µg/mL
-

Immunocytochemistry (ICC/IF)

1-4 µg/mL
-

Flow Cytometry (Flow)

1-2 µg/10^6 cells
-
Product Specifications

Species Reactivity

Human, Mouse, Non-human primate

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

LN-2

Immunogen

SU-DHL-4 lymphoma cells
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C, do not freeze

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Does not react with Rat

Target Information

HLA-DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of a 36 kDa alpha chain (DRA) and 27 kDa beta chain (DRB). The alpha chain gene contains 5 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, and exon 4 encodes the transmembrane domain and the cytoplasmic tail. DRA does not have polymorphisms in the peptide binding part and acts as the sole alpha chain for DRB1, DRB3, DRB4 and DRB5. Within the DR molecule the beta chain contains all the polymorphisms specifying the peptide binding specificities. Hundreds of DRB1 alleles have been described and typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. HLA-DR is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides, present them to T-helper cells, and have a critical role in the initiation of the immune response.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD74; CD74 antigen; CD74 antigen (invariant polypeptide of major histocompatibility complex, class II antigen-associated); CD74 molecule, major histocompatibility complex, class II invariant chain; class II-associated invariant chain peptide; dinucleotide microsatellite; gamma chain of class II antigens; H-2 class II histocompatibility antigen gamma chain; histocompatibility: class II antigens, gamma chain of; HLA class II histocompatibility antigen gamma chain; HLA-DR antigens-associated invariant chain; HLA-DR-GAMMA; Ia antigen-associated invariant chain; Ia-associated invariant chain; Ii; invariant polypeptide of major histocompatibility complex, class II antigen-associated; MHC class II-associated invariant chain; MHC HLA-DR gamma chain; p33

View more View less

Gene Aliases: CD74; CLIP; DHLAG; HLADG; Ia-GAMMA; II

View more View less

UniProt ID: (Human) P04233, (Mouse) P04441

View more View less

Entrez Gene ID: (Human) 972, (Mouse) 16149

View more View less

Function(s)
beta-amyloid binding cytokine receptor activity protein binding cytokine binding MHC class II protein complex binding macrophage migration inhibitory factor binding MHC class II protein binding MHC class II protein binding, via antigen binding groove identical protein binding protein binding involved in protein folding nitric-oxide synthase binding
Process(es)
activation of MAPK activity prostaglandin biosynthetic process positive regulation of cytokine-mediated signaling pathway immune system process positive regulation of dendritic cell antigen processing and presentation negative regulation of peptide secretion positive regulation of adaptive immune response positive regulation of type 2 immune response negative regulation of mature B cell apoptotic process protein folding protein complex assembly intracellular protein transport defense response immune response signal transduction cell proliferation immunoglobulin mediated immune response cytokine-mediated signaling pathway antigen processing and presentation antigen processing and presentation of exogenous peptide antigen via MHC class II positive regulation of B cell proliferation macrophage migration inhibitory factor signaling pathway negative regulation of apoptotic process negative regulation of DNA damage response, signal transduction by p53 class mediator positive thymic T cell selection negative thymic T cell selection positive regulation of innate immune response negative regulation of T cell differentiation positive regulation of T cell differentiation positive regulation of fibroblast proliferation positive regulation of peptidyl-tyrosine phosphorylation chaperone mediated protein folding requiring cofactor positive regulation of macrophage cytokine production positive regulation of ERK1 and ERK2 cascade positive regulation of neutrophil chemotaxis negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator positive regulation of chemokine (C-X-C motif) ligand 2 production antigen processing and presentation of endogenous antigen regulation of macrophage activation T cell selection leukocyte migration
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0