Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD70 Antibodies

Leinco Technologies

CD70 Monoclonal Antibody (TAN1-7), Functional Grade, In Vivo GOLD™

View all (46) CD70 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD70 Monoclonal Antibody (TAN1-7), Functional Grade, In Vivo GOLD™

CD70 Monoclonal Antibody (TAN1-7), Functional Grade, In Vivo GOLD™

Product Details

I-2002-100MG

Applications
Tested Dilution
Publications

Immunocytochemistry (ICC/IF)

Assay-dependent
-

Flow Cytometry (Flow)

Assay-dependent
-

ELISA (ELISA)

Assay-dependent
-

Functional Assay (Functional)

Assay-dependent
-

Inhibition Assays (IA)

Assay-dependent
-

Blocking Assay (BLOCK)

Assay-dependent
-
Product Specifications

Species Reactivity

Mouse

Host/Isotype

Mouse / IgG2a, kappa

Class

Monoclonal

Type

Antibody

Clone

TAN1-7

Immunogen

Recombinant mouse CD70 (extracellular domain, aa residues 41-195)
View immunogen

Conjugate

Functional Grade Functional Grade Functional Grade

Form

Liquid

Concentration

≥5.0 mg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.2-7.4

Contains

no preservative

Storage conditions

4°C short term, -80°C long term

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Specificity: Clone TAN1-7 monoclonal antibody recognizes a direct epitope on mouse CD70 and capable of blocking the CD70-CD27 interaction involved in B and T cell activation.

Endotoxin Level: <1.0 EU/mg as determined by the LAL method.

Purity: ≥95% monomer by analytical SEC and >95% by SDS Page.

Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.

Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.

Target Information

The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T and B lymphocytes. It induces proliferation of costimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD27 ligand; CD27-L; CD70; CD70 antigen; Ki-24 antigen; tumor necrosis factor (ligand) superfamily, member 7; Tumor necrosis factor ligand superfamily member 7

View more View less

Gene Aliases: Cd27l; CD27LG; Cd70; Tnfsf7

View more View less

UniProt ID: (Mouse) O55237

View more View less

Entrez Gene ID: (Mouse) 21948

View more View less

Function(s)
protease binding receptor binding cytokine activity tumor necrosis factor receptor binding
Process(es)
immune response signal transduction cell-cell signaling cell proliferation extrinsic apoptotic signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0