Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD70 Antibodies

Invitrogen

CD70 Monoclonal Antibody (BU69), Biotin

View all (46) CD70 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD70 Monoclonal Antibody (BU69), Biotin

CD70 Monoclonal Antibody (BU69), Biotin

Product Details

MA5-17726

Applications
Tested Dilution
Publications

Immunohistochemistry (Frozen) (IHC (F))

Assay-dependent
-

Flow Cytometry (Flow)

Assay-dependent
-

Immunoprecipitation (IP)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

BU69

Immunogen

Waldenstroms macroglobulineamia cell line WM-1.
View immunogen

Conjugate

Biotin Biotin Biotin
  • Unconjugated
  • FITC
  • PE 
  • Request custom conjugation

Form

Liquid

Concentration

0.1 mg/mL

Purification

Protein G

Storage buffer

PBS with 4-5mg/mL BSA

Contains

0.02% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2539116

Product Specific Information

MA5-17726 targets CD70/Ki-24 antigen in FACS, IHC (F), and IP applications and shows reactivity with Human samples.

The MA5-17726 immunogen is mouse monoclonal antibodies raised against human CD70.

Target Information

The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T and B lymphocytes. It induces proliferation of costimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD27 ligand; CD27-L; CD70; CD70 antigen; Ki-24 antigen; surface antigen CD70; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8A; Tumor necrosis factor ligand superfamily member 7

View more View less

Gene Aliases: CD27L; CD27LG; CD70; TNFSF7; TNLG8A

View more View less

UniProt ID: (Human) P32970

View more View less

Entrez Gene ID: (Human) 970

View more View less

Function(s)
protease binding receptor binding cytokine activity tumor necrosis factor receptor binding protein binding
Process(es)
immune response signal transduction cell-cell signaling cell proliferation tumor necrosis factor-mediated signaling pathway extrinsic apoptotic signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0