Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD70 Antibodies

Invitrogen

CD70 Recombinant Rabbit Monoclonal Antibody (BLR176J)

View all (46) CD70 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD70 Recombinant Rabbit Monoclonal Antibody (BLR176J)

CD70 Recombinant Rabbit Monoclonal Antibody (BLR176J)

Product Details

MA5-44537

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

BLR176J

Immunogen

Recombinant protein representing the extracellular domain of Human CD70 (residues 50-193).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

100 µg/mL

Purification

purified

Storage buffer

BBS, pH 7.8-8.6, with 0.1% BSA

Contains

0.09% sodium azide

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2926667

Target Information

The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T and B lymphocytes. It induces proliferation of costimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD27 ligand; CD27-L; CD70; CD70 antigen; Ki-24 antigen; surface antigen CD70; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8A; Tumor necrosis factor ligand superfamily member 7

View more View less

Gene Aliases: CD27L; CD27LG; CD70; TNFSF7; TNLG8A

View more View less

UniProt ID: (Human) P32970

View more View less

Entrez Gene ID: (Human) 970

View more View less

Function(s)
protease binding receptor binding cytokine activity tumor necrosis factor receptor binding protein binding
Process(es)
immune response signal transduction cell-cell signaling cell proliferation tumor necrosis factor-mediated signaling pathway extrinsic apoptotic signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0