Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • E-selectin (CD62E) Antibodies

Invitrogen

CD62E (E-selectin) Monoclonal Antibody (CL-37), eBioscience™

View all (42) E-selectin (CD62E) antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD62E (E-selectin) Monoclonal Antibody (CL-37), eBioscience™

Product Details

BMS1014

Applications
Tested Dilution
Publications

Immunohistochemistry (Frozen) (IHC (F))

Assay-Dependent
-

ELISA (ELISA)

Assay-Dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

CL-37

Immunogen

Rec. Human CD62E (E-selectin)

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2-7.4

Contains

no preservative

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_10596513

Product Specific Information

Description: E-Selectin (CL-37) is a mouse monoclonal antibody raised against E-Selectinof human origin.

Endothelial Leukocyte Adhesion Molecule-1 belongs to the selectin family of adhesion molecules. Together with L-selectin and P-selectin, E-selectin mediates the initial interactions of leukocytes and platelets with endothelial cells.

Molecular structure: The extracellular part of all selectins consists of an aminoterminal c-type lectin domain which specifically binds to carbohydrate ligands. This is followed by an EGF-like domain, and, in the case of E-selectin, by 6 short consensus repeats. The transmembrane portion of the molecule is followed by a short cytoplasmic tail.

Selectins guide non-activated polymorphonuclear cells to the areas of inflammation in creating first, loose contacts with the endothelial layer. Together with P-selectin, E-selectin is expressed on cytokine-activated endothelial cells, and contributes to the adhesion of still resting leukocytes to the endothelium. This initial binding event is an essential prerequisite for the activation of the immune cells via different inflammatory mediators. In contrast to P-selectin, E-selectin is maximally expressed 2-4 hours after cell activation. Within the next 24-48 hours E-selectin is again eliminated from the cytoplasmic membrane by shedding into the circulation.

Applications Tested: ELISA, Immunohistochemistry (Frozen).

Source: Plant expressed 130 amino acids (His25-Ser153) with 16 amino acid Histidine tag. accession number NP_000580.

Bioactivity: The ED50 of this protein, as measured by TF-1 cell proliferation assay, is less than or equal to 0.7 ng/mL. This corresponds to a specific activity of greater than or equal to 5 x 10e6 Units/mg.

Endotoxin: Less than 0.005 ng/ug (0.05 EU/ug) as measured by turbidity assay. Purity: Greater than or equal to 95% as determined by SDS-PAGE.

Molecular Weight: 17.2 kDa (apparent 23 and 24 kDa).

Target Information

Leukocytes adhere to the blood vessel endothelium during extravasation in postcapillary venules of lymph nodes. In addition, leukocyte adhesion occurs in the capillaries and small venules at any site in the body after onset of inflammation. This response is immediate and involves a cascade of adhesion receptors. At the endothelial surface members of the selectin and immunoglobulin superfamilies participate in this cascade. Selectins are C-type cell surface lectins that play a role in leukocyte adhesion to the blood vessel wall endothelium. E-selectin (CD62E) is an endothelial cell specific selectin that is expressed only after activation with proinflammatory cytokines. In vitro experiments have shown that IL-1, TNFalpha and bacterial wall components like lipopolysaccharides induce the transcription of E-selectin in a NFkB dependent signaling cascade. E-selectin has been associated with blood vessel endothelium in diverse inflammatory situations.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD62 antigen-like family member E; CD62E; e selectin; E-selectin; ELAM-1; endothelial adhesion molecule 1; Endothelial leukocyte adhesion molecule 1; LECAM2; leukocyte endothelial cell adhesion molecule 2; Leukocyte-endothelial cell adhesion molecule 2; RP1-117P20.2

View more View less

Gene Aliases: CD62E; ELAM; ELAM1; ESEL; LECAM2; SELE

View more View less

UniProt ID: (Human) P16581

View more View less

Entrez Gene ID: (Human) 6401

View more View less

Function(s)
transmembrane signaling receptor activity protein binding sialic acid binding phospholipase binding oligosaccharide binding
Process(es)
positive regulation of receptor internalization leukocyte migration involved in inflammatory response inflammatory response heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules leukocyte cell-cell adhesion activation of phospholipase C activity calcium-mediated signaling actin filament-based process response to lipopolysaccharide response to tumor necrosis factor regulation of inflammatory response leukocyte migration leukocyte tethering or rolling response to interleukin-1
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0