Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD33 Antibodies

Invitrogen

CD33 Recombinant Rabbit Monoclonal Antibody (001), APC

View all (149) CD33 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD33 Recombinant Rabbit Monoclonal Antibody (001), APC

  • Antibody Testing Data (1)
CD33 Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
CD33 Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD33 Antibody (MA5-46746) in Flow

Flow cytometry analysis of CD33 in human whole blood monocytes (left panel) and granulocytes (right panel) using CD33, APC Recombinant Monoclonal (Product # MA5-46746). The histograms were derived from gated events with the forward and side light-scatter characteristics of viable monocytes and granulocytes. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD33 Antibody in Flow Cytometry (Flow)
CD33 Recombinant Rabbit Monoclonal Antibody (001), APC

Product Details

MA5-46746

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

5 µL/test
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

001

Immunogen

Recombinant protein ecoding Human CD33 (Met1-His259).
View immunogen

Conjugate

APC APC APC
  • Unconjugated
  • FITC
  • PE 
  • PerCP 
  • Request custom conjugation

Excitation/Emission Max

651/660 nm View spectra spectra

Form

Liquid

Concentration

100 µg/mL

Purification

Protein A

Storage buffer

PBS with 0.5% BSA

Contains

0.03% Proclin 300

Storage conditions

4°C, store in dark, DO NOT FREEZE!

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2937818

Product Specific Information

Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human CD33 (Met1-His259) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze!

Target Information

CD33 is a transmembrane protein of the sialic acid-binding immunoglobulin-like lectin (Siglec) family. It belongs to the immunoreceptor tyrosine-based inhibitory motif (ITIM)-containing molecules able of recruiting protein tyrosine phosphatases SHP-1 and SHP-2 to signal assemblies, and these ITIMs are also used for ubiquitin-mediated removal of the receptor from the cell surface. CD33 is expressed on cells of myelomonocytic lineage, binds sialic acid residues in N- and O-glycans on cell surfaces, and is a therapeutic target for acute myeloid leukemia. Further, CD33 is found on granulocyte and macrophage precursors in the bone marrow, but is not on pluripotent stem cells. CD33 is also expressed on, and is a useful marker for, peripheral monocytes. CD33 is useful for distinguishing myelogenous leukemia cells from lymphoid or erythroid leukemias. Diseases associated with CD43 dysfunction include gallbladder lymphoma and extracutaneous mastocytoma.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD33; CD33 antigen (gp67); CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; sialic acid binding Ig-like lectin 3; Sialic acid-binding Ig-like lectin 3; Siglec-3

View more View less

Gene Aliases: CD33; p67; SIGLEC-3; SIGLEC3

View more View less

UniProt ID: (Human) P20138

View more View less

Entrez Gene ID: (Human) 945

View more View less

Function(s)
receptor activity protein binding carbohydrate binding
Process(es)
cell adhesion signal transduction cell-cell signaling negative regulation of cell proliferation regulation of immune response
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0