Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • JAM-A (CD321) Antibodies

Invitrogen

CD321 (F11R) Monoclonal Antibody (H202-106)

View all (36) JAM-A (CD321) antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD321 (F11R) Monoclonal Antibody (H202-106)

  • Antibody Testing Data (1)
CD321 (F11R) Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
CD321 (F11R) Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD321 (F11R) Antibody (MA5-28179) in Flow

Flow cytometry of CD321 (F11R) in mouse peripheral blood platelets. Sample was incubated with CD321 (F11R) monoclonal antibody (Product # MA5-28179). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD321 (F11R) Antibody in Flow Cytometry (Flow)
CD321 (F11R) Monoclonal Antibody (H202-106)

Product Details

MA5-28179

Applications
Tested Dilution
Publications

Immunohistochemistry (Frozen) (IHC (F))

Assay-dependent
-

Flow Cytometry (Flow)

Assay-dependent
-

Immunoprecipitation (IP)

Assay-dependent
-
Product Specifications

Species Reactivity

Mouse

Host/Isotype

Rat / IgG1

Class

Monoclonal

Type

Antibody

Clone

H202-106

Immunogen

MTE1/MTE2 stromal cell lines
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated
  • Biotin 
  • Request custom conjugation

Form

Liquid

Concentration

1 mg/mL

Purification

Protein G

Storage buffer

PBS

Contains

0.09% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2745155

Product Specific Information

This antibody recognizes murine CD321, also known as junctional adhesion molecule 1 (JAM-1). CD321 is a 274 amino acid, single pass, type I transmembrane glycoprotein, which shares similarities with related proteins JAM-2 and JAM-3. CD321 is a multifunctional protein primarily expressed by platelets, endothelial and epithelial cells.

Target Information

CD321 (F11R, JAM-1, JAM-A) is a cell-surface adhesion molecule belonging to the Ig superfamily of receptors. CD321 is a 32-41 kD glycoprotein, possessing two extracellular Ig-like domains, a transmembrane region, and a short cytosolic tail. The protein assembles at the membrane as a homodimer, and can associate laterally with other membrane proteins including integrins. CD321 is expressed on a variety of cell types, including leukocytes, platelets, erythrocytes, stem cells, epithelial and endothelial cells, and plays a role in mediating cell-cell contact. CD321 can be found in close proximity to several components of epithelial tight junctions, including ZO-1 and occludin. CD321 is involved in the maintenance of epithelial and endothelial barriers, hematopoiesis, angiogenesis, and tumor proliferation, invasion, and metastasis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: BV11 antigen; CD321; F11 R; F11 receptor/F11R; JAM-1; JAM-A; junction cell adhesion molecule A; junction cell adhesion molecule1; Junctional adhesion molecule 1; Junctional adhesion molecule A

View more View less

Gene Aliases: 9130004G24; AA638916; ESTM33; F11r; JAM; JAM-1; JAM-A; Jam1; Jcam; Jcam1; Ly106

View more View less

UniProt ID: (Mouse) O88792

View more View less

Entrez Gene ID: (Mouse) 16456

View more View less

Function(s)
protein binding PDZ domain binding
Process(es)
cell adhesion viral process epithelial cell differentiation actomyosin structure organization negative regulation of GTPase activity positive regulation of GTPase activity positive regulation of blood pressure intestinal absorption establishment of protein localization to plasma membrane establishment of endothelial intestinal barrier regulation of membrane permeability regulation of actin cytoskeleton reorganization
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-c7d89468c-sbmwt:80/100.66.128.142:80.
git-commit: 4db89d68e0ee702f860d53b64def1eb94897fade
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0