Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD28 Antibodies

NeoBiotechnologies

CD28 Monoclonal Antibody (C28/77)

View all (167) CD28 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD28 Monoclonal Antibody (C28/77)

  • Antibody Testing Data (1)
CD28 Antibody in SDS-PAGE (SDS-PAGE)
Group 53 Created with Sketch.
CD28 Antibody in SDS-PAGE (SDS-PAGE)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD28 Antibody (940-MSM3-P1ABX) in SDS-PAGE

SDS-PAGE analysis Purified CD28 Mouse Monoclonal Antibody (C28/77). Confirmation of Integrity and Purity of Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD28 Antibody in SDS-PAGE (SDS-PAGE)

Product Details

940-MSM3-P1ABX

Applications
Tested Dilution
Publications

ELISA (ELISA)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

C28/77

Immunogen

Recombinant human CD28 protein

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

CD28 antigen is a 44 kDa disulfide linked homodimeric T cell specific surface glycoprotein. CD28 is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes. Moreover, CD28 is the critical T cell costimulatory receptor that provides the cell the important second activation signal by binding CD80 and CD86 which are expressed by antigen presenting cells. In addition to its co-stimulation role, CD28 functions by preventing T cells from entering an anergic-hyporesponsive state or from undergoing premature apoptotic cell death. In murine peripheral lymphoid organs and in the blood, all CD4+ and CD8+ T cells express CD28. In the thymus, CD28 expression is highest on immature CD3-, CD8+ and CD4+8+ cells, and on CD4-8- cells that express alpha/beta and gamma/delta TCR. The level of CD28 on mature CD4+ and CD8+ alpha/beta TCR+ thymocytes is two- to fourfold lower than on the immature cells. Diseases associated with CD28 dysfunction include mycosis fungiodes and Sezary's Disease.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD28; CD28 antigen; MGC138290; sCD28; soluble CD28; T-cell-specific surface glycoprotein CD28; T44; TP44

View more View less

Gene Aliases: CD28; Tp44

View more View less

UniProt ID: (Human) P10747

View more View less

Entrez Gene ID: (Human) 940

View more View less

Function(s)
protease binding SH3/SH2 adaptor activity protein binding coreceptor activity protein kinase binding identical protein binding phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Process(es)
positive regulation of inflammatory response to antigenic stimulus humoral immune response cell surface receptor signaling pathway positive regulation of gene expression negative regulation of gene expression regulation of phosphatidylinositol 3-kinase signaling positive regulation of phosphatidylinositol 3-kinase signaling T cell costimulation positive regulation of interleukin-10 production positive regulation of interleukin-4 production cytokine biosynthetic process positive regulation of T cell proliferation negative thymic T cell selection regulatory T cell differentiation positive regulation of viral genome replication positive regulation of interleukin-2 biosynthetic process regulation of regulatory T cell differentiation positive regulation of translation positive regulation of mitotic nuclear division positive regulation of transcription from RNA polymerase II promoter positive regulation of alpha-beta T cell proliferation phosphatidylinositol phosphorylation phosphatidylinositol-mediated signaling positive regulation of isotype switching to IgG isotypes regulation of defense response to virus by virus T cell receptor signaling pathway positive regulation of protein kinase B signaling apoptotic signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0