Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • B7-H3 (CD276) Antibodies

Abnova

CD276 Monoclonal Antibody (7G6)

View all (92) B7-H3 (CD276) antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD276 Monoclonal Antibody (7G6)

  • Antibody Testing Data (1)
CD276 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
CD276 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD276 Antibody (H00080381-M11) in ICC/IF

Immunofluorescence of monoclonal antibody to CD276 on MCF-7 cell. Antibody concentration is 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD276 Antibody in Immunocytochemistry (ICC/IF)
CD276 Monoclonal Antibody (7G6)

Product Details

H00080381-M11

Applications
Tested Dilution
Publications

Immunocytochemistry (ICC/IF)

10 µg/mL
-

ELISA (ELISA)

10 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

7G6

Immunogen

CD276 (AAH62581.1, 28 a.a. approximately 238 a.a) partial recombinant protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Sequence of this protein is as follows: ALEVQVPEDP VVALVGTDAT LCCSFSPEPG FSLAQLNLIW QLTDTKQLVH SFAEGQDQGS AYANRTALFP DLLAQGNASL RLQRVRVADE GSFTCFVSIR DFGSAAVSLQ VAAPYSKPSM TLEPNKDLRP GDTVTITCSS YQGYPEAEVF WQDGQGVPLT GNVTTSQMAN EQGLFDVHSI LRVVLGANGT YSCLVRNPVL QQDAHSSVTI T

Target Information

CD276 is a member of the B7 family of co-stimulatory molecules also known as B7-H3. CD276 is a type I transmembrane protein that induces the proliferation of CD4+ and CD8+ T cells, enhances the generation of cytotoxic T cells and selectively stimulates the production of interferon gamma. Expression of CD276 can be induced on dendritic cells and monocytes by inflammatory cytokines, and is also widely expressed in peripheral tissues including the heart, kidney, testis and colon. In humans, CD276 exists as two isoforms which result from gene duplication and differential splicing. CD276 is reported to have therapeutic potential for the regulation of cell-mediated immune responses to cancer, particularly in conjunction with anti-angiogenic therapy.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 4Ig-B7-H3; B7 homolog 3; B7-H3; CD276; CD276 antigen; Costimulatory molecule; sB7 H3; sB7H3; sCD276; soluble B7 H3; soluble B7H3; soluble CD276

View more View less

Gene Aliases: 4Ig-B7-H3; B7-H3; B7H3; B7RP-2; CD276; PSEC0249; UNQ309/PRO352

View more View less

UniProt ID: (Human) Q5ZPR3

View more View less

Entrez Gene ID: (Human) 80381

View more View less

Function(s)
receptor binding protein binding
Process(es)
immune response cell proliferation positive regulation of bone mineralization positive regulation of T cell proliferation T cell activation negative regulation of T cell proliferation negative regulation of interferon-gamma biosynthetic process positive regulation of interferon-gamma biosynthetic process negative regulation of interleukin-2 biosynthetic process positive regulation of osteoblast differentiation negative regulation of inflammatory response regulation of immune response positive regulation of interleukin-2 secretion
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0