Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • BAFF (BLyS) Antibodies

NeoBiotechnologies

CD257/BAFF/TNFSF13B Monoclonal Antibody (C257/1638)

View all (33) BAFF (BLyS) antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD257/BAFF/TNFSF13B Monoclonal Antibody (C257/1638)

  • Antibody Testing Data (1)
CD257/BAFF/TNFSF13B Antibody in SDS-PAGE (SDS-PAGE)
Group 53 Created with Sketch.
CD257/BAFF/TNFSF13B Antibody in SDS-PAGE (SDS-PAGE)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD257/BAFF/TNFSF13B Antibody (10673-MSM1-P0) in SDS-PAGE

SDS-PAGE analysis Purified CD257 Mouse Monoclonal Antibody (C257/1638). Confirmation of Purity and Integrity of Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD257/BAFF/TNFSF13B Antibody in SDS-PAGE (SDS-PAGE)
CD257/BAFF/TNFSF13B Monoclonal Antibody (C257/1638)

Product Details

10673-MSM1-P0

Applications
Tested Dilution
Publications

ELISA (ELISA)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

C257/1638

Immunogen

Purified recombinant human CD257 protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C, do not freeze

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

BAFF is a membrane bound cytokine expressed by B cell lineage cells. The soluble form of the cytokine binds to the receptors TNFRSF13B/TACI, TNFRSF17/BCMA, and TNFRSF13C/BAFFR. BAFF is a potent B cell activator, and important for the proliferation and differentiation of B cells. BAFF is also expressed by T lymphocytes and has been shown to promote T cell activation and survival. Isoforms representing alternatively spliced, and glycosylated variants have been identified.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: ApoL related ligand TALL-1; B lymphocyte stimulator; B-cell-activating factor; B-lymphocyte stimulator; BAFF; BLyS; CD257; delta BAFF; Delta4 BAFF; Dendritic cell-derived TNF-like molecule; TALL-1; TNF and ApoL-related leukocyte expressed ligand 1; TNF homolog that activates apoptosis; TNF- and APOL-related leukocyte expressed ligand 1; tumor necrosis factor (ligand) superfamily member 13b; tumor necrosis factor (ligand) superfamily, member 13b; tumor necrosis factor (ligand) superfamily, member 20; tumor necrosis factor ligand 7A; Tumor necrosis factor ligand superfamily member 13B; tumor necrosis factor-like protein ZTNF4

View more View less

Gene Aliases: BAFF; BLYS; CD257; DTL; TALL-1; TALL1; THANK; TNFSF13B; TNFSF20; TNLG7A; UNQ401/PRO738; ZTNF4

View more View less

UniProt ID: (Human) Q9Y275

View more View less

Entrez Gene ID: (Human) 10673

View more View less

Function(s)
receptor binding cytokine activity tumor necrosis factor receptor binding protein binding
Process(es)
immune response signal transduction cell proliferation positive regulation of cell proliferation positive regulation of B cell proliferation tumor necrosis factor-mediated signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0