Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CCL23 Antibodies

NeoBiotechnologies

CCL23/Myeloid Progenitor Inhibitory Factor 1 Monoclonal Antibody (CCL23/4036)

View all (15) CCL23 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CCL23/Myeloid Progenitor Inhibitory Factor 1 Monoclonal Antibody (CCL23/4036)

  • Antibody Testing Data (1)
CCL23/Myeloid Progenitor Inhibitory Factor 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
CCL23/Myeloid Progenitor Inhibitory Factor 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CCL23/Myeloid Progenitor Inhibitory Factor 1 Antibody (6368-MSM6-P1) in IHC (P)

Formalin-fixed, paraffin-embedded human adrenal gland stained with CCL23 Mouse Monoclonal Antibody (CCL23/4036). HIER: Tris/EDTA, pH9.0, 45 min. 2°C: HRP-polymer, 30 min. DAB, 5 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CCL23/Myeloid Progenitor Inhibitory Factor 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CCL23/Myeloid Progenitor Inhibitory Factor 1 Monoclonal Antibody (CCL23/4036)

Product Details

6368-MSM6-P1

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1-2 µg/mL
-

Peptide Array (Array)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2c, kappa

Class

Monoclonal

Type

Antibody

Clone

CCL23/4036

Immunogen

Recombinant fragment (around aa1-120) of human CCL23 protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Positive Control: High levels in adult lung liver skeletal muscle and pancreas.

Cellular Location: Secreted.

Target Information

This gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. This chemokine, a member of the CC subfamily, displays chemotactic activity on resting T lymphocytes and monocytes, lower activity on neutrophils and no activity on activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. In addition, the product of this gene is a potent agonist of the chemokine (C-C motif) receptor 1. Alternative splicing results in multiple transcript variants that encode different isoforms.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: C-C motif chemokine 23; C6 beta-chemokine; chemokine (C-C motif) ligand 23; Ck beta 8-1; CK-beta-8; CKB-8; Macrophage inflammatory protein 3; MIP-3; MIP-3CK-BETA-8; MPIF-1; MPIF-1Ckb-8-1; Myeloid progenitor inhibitory factor 1; small inducible cytokine subfamily A (Cys-Cys), member 23; Small-inducible cytokine A23

View more View less

Gene Aliases: CCL23; CK-BETA-8; Ckb-8; Ckb-8-1; CKb8; hmrp-2a; MIP-3; MIP3; MPIF-1; MPIF1; SCYA23

View more View less

UniProt ID: (Human) P55773

View more View less

Entrez Gene ID: (Human) 6368

View more View less

Function(s)
chemokine activity heparin binding CCR1 chemokine receptor binding
Process(es)
monocyte chemotaxis cellular calcium ion homeostasis chemotaxis inflammatory response immune response signal transduction G-protein coupled receptor signaling pathway cell-cell signaling negative regulation of cell proliferation neutrophil chemotaxis positive regulation of GTPase activity lymphocyte chemotaxis positive regulation of inflammatory response chemokine-mediated signaling pathway positive regulation of ERK1 and ERK2 cascade cellular response to interferon-gamma cellular response to interleukin-1 cellular response to tumor necrosis factor negative regulation of C-C chemokine binding
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0