Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CAPZB Antibodies

Proteintech

CAPZB Monoclonal Antibody (1F9G2)

View all (14) CAPZB antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CAPZB Monoclonal Antibody (1F9G2)

  • Antibody Testing Data (4)
CAPZB Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
CAPZB Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CAPZB Antibody (68190-1-IG) in ICC/IF

Immunofluorescent analysis of (-20°C Ethanol) fixed A431 cells using CAPZB antibody (68190-1-Ig, Clone: 1F9G2 ) at dilution of 1:2000 and CoraLite®488-Conjugated AffiniPure Goat Anti-Mouse IgG(H+L), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CAPZB Antibody in Immunocytochemistry (ICC/IF)
CAPZB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CAPZB Antibody in Western Blot (WB)
CAPZB Antibody in Flow Cytometry (Flow)
CAPZB Monoclonal Antibody (1F9G2)

Product Details

68190-1-IG

Applications
Tested Dilution
Publications

Western Blot (WB)

1:5,000-1:50,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:1,000-1:4,000
-

Immunocytochemistry (ICC/IF)

1:1,000-1:4,000
-

Flow Cytometry (Flow)

0.4 µg/1x10^6 cell suspension
-

ELISA (ELISA)

Assay-dependent
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Mouse / IgG2a

Class

Monoclonal

Type

Antibody

Clone

1F9G2

Immunogen

Recombinant protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated
  • CoraLite Plus 488
  • Request custom conjugation

Form

Liquid

Purification

Protein A

Storage buffer

PBS with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

Product Specific Information

Immunogen sequence: SDQQLDCALD LMRRLPPQQI EKNLSDLIDL VPSLCEDLLS SVDQPLKIAR DKVVGKDYLL CDYNRDGDSY RSPWSNKYDP PLEDGAMPSA RLRKLEVEAN NAFDQYRDLY FEGGVSSVYL WDLDHGFAGV ILIKKAGDGS KKIKGCWDSI HVVEVQEKSS GRTAHYKLTS TVMLWLQTNK SGSGTMNLGG SLTRQMEKDE TVSDCSPHIA NIGRLVEDME NKIRSTLNEI YFGKTKDIVN GL

Target Information

CAPZB is a member of the F-actin capping protein family. This gene encodes the beta subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Cap Z; capping protein (actin filament) muscle Z-line, beta; capping protein beta 1; CapZ beta; F-actin capping protein beta subunit; F-actin-capping protein subunit beta; MGC104401; MGC129749; MGC129750; RP4-657E11.7

View more View less

Gene Aliases: 1700120C01Rik; AI325129; CAPB; CAPPB; Cappb1; CAPZ; CAPZB; CPB1; CPB2; CPbeat2; CPbeta1; CPbeta2

View more View less

UniProt ID: (Human) P47756, (Rat) Q5XI32, (Mouse) P47757

View more View less

Entrez Gene ID: (Human) 832, (Rat) 298584, (Mouse) 12345

View more View less

Function(s)
actin binding actin filament binding cadherin binding involved in cell-cell adhesion molecular_function protein binding beta-tubulin binding
Process(es)
cell morphogenesis cytoskeleton organization biological_process regulation of lamellipodium assembly regulation of cell morphogenesis cell projection organization lamellipodium assembly actin cytoskeleton organization negative regulation of microtubule polymerization neuron projection development muscle fiber development barbed-end actin filament capping negative regulation of filopodium assembly regulation of protein kinase C signaling movement of cell or subcellular component blood coagulation cell-cell adhesion actin filament capping
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0