Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Azurocidin Antibodies

Osenses

CAP37 Polyclonal Antibody

2 References
View all (14) Azurocidin antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CAP37 Polyclonal Antibody

CAP37 Polyclonal Antibody

Product Details

OSA00193W-100UL

Applications
Tested Dilution
Publications

Western Blot (WB)

1:200-1:2,000
-

Immunohistochemistry (IHC)

1:200-1:2,000
-

Miscellaneous PubMed (MISC)

-
View 2 publications 2 publications
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / Ig

Class

Polyclonal

Type

Antibody

Immunogen

A synthetic peptide from aa region 20-60 of human Azurocidin conjugated to an immunogenic carrier protein has been used as the antigen
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

Conc. Not Determined

Storage buffer

whole serum

Contains

no preservative

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.

Specificity of this antibody: CAP37.

Target Information

Azurocidin, also called heparin-binding protein (HBP), is an inactive homolog of serine proteinases residing in granulocytes. Initially it was thought to have only activity against gram-negative bacteria. Later it was also found to act against gram-positive bacteria and C. albicans. Azurocidin is considered a family member of polymorphonuclear leukocytes- derived antimicrobial proteins like defensins, LL-37 and lysozyme. Azurocidin has been recognized as a player in the activation and modulation of the immune response and may act to alarm the immune system. The cationic domain at one side of the protein is essential for its antimicrobial activity. It is stored in azurophil granules as well as secretory granules and as a result partly released at early stage of extravasation. Azurocidin is involved in chemotaxis and activation of monocytes, cytokine release and phagocytosis leading to more efficient bacterial clearance. The primary targeted environment of azurocidin are cells in the bloodstream, that is the endothelial lining, and the extravascular surroundings. The interaction of azurocidin with leucocytes is mediated via beta 2-intergrins. High plasma levels of azurocidin might help with the identification of patients which are at risk in developing sepsis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: Azurocidin; cationic antimicrobial protein 37; Cationic antimicrobial protein CAP37; HBP; Heparin-binding protein; neutrophil azurocidin

View more View less

Gene Aliases: AZAMP; AZU; AZU1; CAP37; HBP; hHBP; HUMAZUR; NAZC

View more View less

UniProt ID: (Human) P20160

View more View less

Entrez Gene ID: (Human) 566

View more View less

Function(s)
serine-type endopeptidase activity protein binding heparin binding peptidase activity toxic substance binding heparan sulfate proteoglycan binding
Process(es)
microglial cell activation proteolysis inflammatory response protein kinase C-activating G-protein coupled receptor signaling pathway glial cell migration positive regulation of gene expression positive regulation of peptidyl-threonine phosphorylation protein processing antimicrobial humoral response calcium-mediated signaling using intracellular calcium source monocyte activation positive regulation of tumor necrosis factor biosynthetic process negative regulation of apoptotic process regulation of vascular permeability cellular extravasation positive regulation of MHC class II biosynthetic process positive regulation of cell adhesion positive regulation of protein kinase activity macrophage chemotaxis positive regulation of interleukin-1 beta biosynthetic process positive regulation of fractalkine biosynthetic process positive regulation of phagocytosis defense response to Gram-negative bacterium induction of positive chemotaxis defense response to virus cell chemotaxis protein kinase C signaling neutrophil mediated killing of bacterium
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0