Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Complement C5a Antibodies

Bioss

C5a anaphylatoxin Polyclonal Antibody, Cyanine5

View all (35) Complement C5a antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite C5a anaphylatoxin Polyclonal Antibody, Cyanine5

C5a anaphylatoxin Polyclonal Antibody, Cyanine5

Product Details

BS-0324R-CY5

Applications
Tested Dilution
Publications

Western Blot (WB)

1:300-5,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-

Immunohistochemistry (Frozen) (IHC (F))

1:50-1:200
-

Immunocytochemistry (ICC/IF)

1:50-1:200
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

KLH conjugated synthetic peptide derived from mouse C5a (725-770aa)
View immunogen

Conjugate

Cyanine5 Cyanine5 Cyanine5

Excitation/Emission Max

647/665 nm View spectra spectra

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

TBS, pH 7.4, with 1% BSA, 50% glycerol

Contains

0.03% ProClin 300

Storage conditions

-20°C or -80°C if preferred

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

The protein encoded by this gene is the fifth component of complement, which plays an important role in inflammatory and cell killing processes. This protein is comprised of alpha and beta polypeptide chains that are linked by a disulfide bridge. An activation peptide, C5a, which is an anaphylatoxin that possesses potent spasmogenic and chemotactic activity, is derived from the alpha polypeptide via cleavage with a convertase. The C5b macromolecular cleavage product can form a complex with the C6 complement component, and this complex is the basis for formation of the membrane attack complex, which includes additional complement components. Mutations in this gene cause complement component 5 deficiency, a disease where patients show a propensity for severe recurrent infections. Defects in this gene have also been linked to a susceptibility to liver fibrosis and to rheumatoid arthritis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: anaphylatoxin C5a analog; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C4B1; C4B12; C4B2; C4B3; C4B5; C4F; C5a anaphylatoxin; CH; CO4; Complement C5; CPAMD3; DADB-112B14.11; FLJ17816; FLJ17822; FLJ60561; Hemolytic complement; MGC142298; MGC164979; prepro-C5

View more View less

Gene Aliases: C5; C5a; C5b; C5D; CPAMD4; ECLZB; Hc; He

View more View less

UniProt ID: (Human) P01031, (Mouse) P06684

View more View less

Entrez Gene ID: (Human) 727, (Mouse) 15139

View more View less

Function(s)
endopeptidase inhibitor activity receptor binding protein binding chemokine activity
Process(es)
activation of MAPK activity in utero embryonic development chemotaxis response to stress inflammatory response complement activation complement activation, alternative pathway complement activation, classical pathway cell surface receptor signaling pathway G-protein coupled receptor signaling pathway positive regulation of vascular endothelial growth factor production negative regulation of macrophage chemotaxis negative regulation of endopeptidase activity cytolysis regulation of complement activation positive regulation of angiogenesis cell chemotaxis positive regulation of chemokine secretion mitophagy in response to mitochondrial depolarization immune system process innate immune response
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0