Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • BXDC5 Antibodies

Invitrogen

BXDC5 Polyclonal Antibody

View all (2) BXDC5 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite BXDC5 Polyclonal Antibody

  • Antibody Testing Data (2)
BXDC5 Antibody in Immunohistochemistry (IHC)
Group 53 Created with Sketch.
BXDC5 Antibody in Immunohistochemistry (IHC)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

BXDC5 Antibody (PA5-41912) in IHC

Immunohistochemistry analysis of human breast tissue using an anti-BXDC5 polyclonal antibody (Product # PA5-41912). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

BXDC5 Antibody in Immunohistochemistry (IHC)
BXDC5 Antibody in Western Blot (WB)
BXDC5 Polyclonal Antibody

Product Details

PA5-41912

Applications
Tested Dilution
Publications

Western Blot (WB)

0.2-1 µg/mL
-

Immunohistochemistry (IHC)

5 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Synthetic peptide directed towards the N-terminal of human BXDC5 (aa 45-94).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.5 mg/mL

Purification

Affinity Chromatography

Storage buffer

PBS with 2% sucrose

Contains

0.09% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2577237

Product Specific Information

Peptide sequence: AAAFPPGFSI SEIKNKQRRH LMFTRWKQQQ RKEKLAAKKK LKKEREALGD

Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 79%

Target Information

BXDC5 may be required for ribosome biogenesis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 2310066N05Rik; brix domain containing 5; Brix domain-containing protein 5; Ribosome biogenesis protein RPF1; Ribosome production factor 1; RNA processing factor 1 (RPF1); RP11-118B23.1

View more View less

Gene Aliases: BXDC5; RPF1

View more View less

UniProt ID: (Human) Q9H9Y2

View more View less

Entrez Gene ID: (Human) 80135

View more View less

Function(s)
RNA binding protein binding rRNA primary transcript binding poly(A) RNA binding
Process(es)
ribosomal large subunit assembly maturation of 5.8S rRNA maturation of LSU-rRNA
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-c7d89468c-sbmwt:80/100.66.128.142:80.
git-commit: 4db89d68e0ee702f860d53b64def1eb94897fade
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0