Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • BRD4 Antibodies

Invitrogen

BRD4 Polyclonal Antibody

2 References
View all (31) BRD4 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite BRD4 Polyclonal Antibody

  • Antibody Testing Data (4)
BRD4 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
BRD4 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

BRD4 Antibody (PA5-41550) in ICC/IF

Immunofluorescence analysis of HeLa cells using an anti-BRD4 polyclonal antibody (Product # PA5-41550). Primary Antibody Dilution: 4 µg/mL; Secondary Antibody: Anti-rabbit Alexa 546; Secondary Antibody Dilution: 2 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

BRD4 Antibody in Immunocytochemistry (ICC/IF)
BRD4 Antibody in Western Blot (WB)
BRD4 Antibody in Western Blot (WB)
BRD4 Antibody in ChIP Assay (CHIP)
BRD4 Polyclonal Antibody

Product Details

PA5-41550

Applications
Tested Dilution
Publications

Western Blot (WB)

0.2-1 µg/mL
-

Immunocytochemistry (ICC/IF)

4 µg/mL
-

ChIP assay (ChIP)

0.5 µg
View 2 publications 2 publications
Product Specifications

Species Reactivity

Human

Published species

Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Synthetic peptide directed towards the C-terminal of human BRD4 (aa 651-700).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.5 mg/mL

Purification

Affinity Chromatography

Storage buffer

PBS with 2% sucrose

Contains

0.09% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2610136

Product Specific Information

Peptide sequence: EIEIDFETLK PSTLRELERY VTSCLRKKRK PQAEKVDVIA GSSKMKGFSS

Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%

Target Information

BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: BRD4-NUT FUSION; bromodomain-containing 4; Bromodomain-containing protein 4; chromosome-associated protein; Protein HUNK1

View more View less

Gene Aliases: BRD4; CAP; HUNK1; HUNKI; MCAP

View more View less

UniProt ID: (Human) O60885

View more View less

Entrez Gene ID: (Human) 23476

View more View less

Function(s)
p53 binding chromatin binding protein binding lysine-acetylated histone binding
Process(es)
regulation of transcription involved in G1/S transition of mitotic cell cycle transcription, DNA-templated cellular response to DNA damage stimulus positive regulation of G2/M transition of mitotic cell cycle viral process covalent chromatin modification positive regulation of transcription elongation from RNA polymerase II promoter positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of transcription from RNA polymerase II promoter regulation of inflammatory response regulation of phosphorylation of RNA polymerase II C-terminal domain
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-657bd577ff-sg76p:80/100.66.128.142:80.
git-commit: cdba60da2f3289684898c31dc50804c008488b19
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0