Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • BFSP2 Antibodies

OriGene

BFSP2 Monoclonal Antibody (OTI4E7), TrueMAB™

View all (28) BFSP2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite BFSP2 Monoclonal Antibody (OTI4E7), TrueMAB™

  • Antibody Testing Data (23)
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 23

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

BFSP2 Antibody (CF804351) in IHC (P)

Immunohistochemical staining of paraffin-embedded human lymphoma tissue using anti-BFSP2 mouse monoclonal antibody. (Heat-induced epitope retrieval by 1 mM EDTA in 10mM Tris, pH8.5, 120°C for 3min, TA804351) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BFSP2 Antibody in Western Blot (WB)
BFSP2 Monoclonal Antibody (OTI4E7), TrueMAB™

Product Details

CF804351

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500
-

Immunohistochemistry (Paraffin) (IHC (P))

1:150
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI4E7

Immunogen

Human recombinant protein fragment corresponding to amino acids 184-415 of human BFSP2 produced in E.coli.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

More than 99% of the vertebrate ocular lens is comprised of terminally differentiated lens fiber cells. Two lens-specific intermediate filament-like proteins, the protein product of this gene (phakinin), and filensin, are expressed only after fiber cell differentiation has begun. Both proteins are found in a structurally unique cytoskeletal element that is referred to as the beaded filament (BF). Mutations in this gene have been associated with juvenile-onset, progressive cataracts and Dowling-Meara epidermolysis bullosa simplex.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 49 kDa cytoskeletal protein; beaded filament protein CP49; Beaded filament structural protein 2; beaded filament structural protein 2, phakinin; bfps2, Cytoskeletal protein, 49-kD; CP47; CP49; Lens fiber cell beaded filament protein CP 47; Lens fiber cell beaded filament protein CP 49; Lens intermediate filament-like light; LIFL-L; MGC142078; MGC142080; Phakinin

View more View less

Gene Aliases: BFSP2; CP47; CP49; CTRCT12; LIFL-L; PHAKOSIN

View more View less

UniProt ID: (Human) Q13515

View more View less

Entrez Gene ID: (Human) 8419

View more View less

Function(s)
structural constituent of cytoskeleton structural constituent of eye lens protein binding
Process(es)
visual perception intermediate filament cytoskeleton organization cell maturation response to stimulus lens fiber cell development
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0