Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • BDNF Antibodies

Proteintech

BDNF Monoclonal Antibody (1E5C5), CoraLite® Plus 488

View all (46) BDNF antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite BDNF Monoclonal Antibody (1E5C5), CoraLite® Plus 488

  • Antibody Testing Data (1)
BDNF Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
BDNF Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

BDNF Antibody (CL488-66292) in ICC/IF

Immunofluorescent analysis of (-20°C Ethanol) fixed PC-12 cells using CL488-66292 (BDNF antibody) at dilution of 1:100. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

BDNF Antibody in Immunocytochemistry (ICC/IF)

Product Details

CL488-66292

Applications
Tested Dilution
Publications

Immunocytochemistry (ICC/IF)

1:50-1:500
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2a

Class

Monoclonal

Type

Antibody

Clone

1E5C5

Immunogen

Recombinant protein

Conjugate

CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
  • Unconjugated
  • CoraLite 594
  • Request custom conjugation

Excitation/Emission Max

493/522 nm

Form

Liquid

Purification

Protein A

Storage buffer

PBS with 0.5% BSA, 50% glycerol

Contains

0.05% ProClin 300

Storage conditions

-20°C, store in dark

Shipping conditions

Wet ice

Product Specific Information

Immunogen sequence: MKEANIRGQG GLAYPGVRTH GTLESVNGPK AGSRGLTSLA DTFEHMIEEL LDEDQKVRPN EENNKDADLY TSRVMLSSQV PLEPPLLFLL EEYKNYLDAA NMSMRVRRHS DPARRGELSV CDSISEWVTA ADKKTAVDMS GGTVTVLEKV PVSKGQLKQY FYETKCNPMG YTKEGCRGID KRHWNSQCRT TQSYVRALTM DSKKRIGWRF IRIDTSCVCT LTIKRGR

Target Information

BDNF (Brain-Derived Neurotrophic Factor) is a member of the neurotrophin family. BDNF is synthesized as pre-proBDNF, followed by cleavage to proBDNF. Although further processing generates the mature, 14 kDa protein, pro-BDNF is biologically active and is secreted in synaptic vesicles along with the mature form. BDNF is widely expressed in the central nervous system and acts in an autocrine and paracrine manner on several classes of neurons. Signaling occurs mainly through the tyrosine kinase receptor TrkB, although binding to the lower-affinity receptor, p75-NTR, has also been demonstrated. BDNF promotes neuronal survival and differentiation and has been shown to play a critical role in memory formation and synaptic regulation. BDNF is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of BDNF is reduced in both Alzheimer's and Huntington disease patients, and may also play a role in the regulation of stress response and in the biology of mood disorders.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Abrineurin; Brain-derived neurotrophic factor; H-BDNF; Neurotrophic factor BDNF precursor form; neurotrophin; proBDNF

View more View less

Gene Aliases: ANON2; BDNF; BULN2

View more View less

UniProt ID: (Human) P23560

View more View less

Entrez Gene ID: (Human) 627

View more View less

Function(s)
neurotrophin TRKB receptor binding growth factor activity
Process(es)
cell-cell signaling nervous system development axon guidance synapse assembly brain-derived neurotrophic factor receptor signaling pathway positive regulation of brain-derived neurotrophic factor receptor signaling pathway negative regulation of neuron apoptotic process collateral sprouting positive regulation of collateral sprouting positive regulation of synapse assembly
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0