Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • BCAT2 Antibodies

Invitrogen

BCAT2 Polyclonal Antibody

View all (12) BCAT2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite BCAT2 Polyclonal Antibody

  • Antibody Testing Data (5)
BCAT2 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
BCAT2 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

BCAT2 Antibody (PA5-121097) in ICC/IF

Immunofluorescence analysis of BCAT2 in NIH-3T3 cells. Samples were incubated with BCAT2 Polyclonal antibody (Product # PA5-121097) using a dilution of 1:100 (40x lens). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

BCAT2 Antibody in Immunocytochemistry (ICC/IF)
BCAT2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BCAT2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
BCAT2 Antibody in Western Blot (WB)
BCAT2 Antibody in Western Blot (WB)
BCAT2 Polyclonal Antibody

Product Details

PA5-121097

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-

Immunocytochemistry (ICC/IF)

1:50-1:200
-

ELISA (ELISA)

1 µg/mL
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 28-392 of human BCAT2 (NP_001181.2).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.73 mg/mL

Purification

Affinity Chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2914669

Product Specific Information

Positive test controls include: MCF7, Jurkat, Mouse brain, Rat kidney. The target is usually found in the following locations: Cytoplasm, Mitochondrion.

Immunogen sequence: ASSSFKAADL QLEMTQKPHK KPGPGEPLVF GKTFTDHMLM VEWNDKGWGQ PRIQPFQNLT LHPASSSLHY SLQLFEGMKA FKGKDQQVRL FRPWLNMDRM LRSAMRLCLP SFDKLELLEC IRRLIEVDKD WVPDAAGTSL YVRPVLIGNE PSLGVSQPTR ALLFVILCPV GAYFPGGSVT PVSLLADPAF IRAWVGGVGN YKLGGNYGPT VLVQQEALKR GCEQVLWLYG PDHQLTEVGT MNIFVYWTHE DGVLELVTPP LNGVILPGVV RQSLLDMAQT WGEFRVVERT ITMKQLLRAL EEGRVREVFG SGTACQVCPV HRILYKDRNL HIPTMENGPE LILRFQKELK EIQYGIRAHE WMFPV

Target Information

This gene encodes a branched chain aminotransferase found in mitochondria. The encoded protein forms a dimer that catalyzes the first step in the production of the branched chain amino acids leucine, isoleucine, and valine. Multiple transcript variants encoding different isoforms have been found for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: BCAT(m); branched chain amino-acid transaminase 2, mitochondrial; branched chain aminotransferase 2, mitochondrial; Branched-chain-amino-acid aminotransferase, mitochondrial; heart branched chain aminotransferase precursor encoding mitochondrial protein; Placental protein 18; PP18

View more View less

Gene Aliases: BCAM; Bcat-2; BCAT2; BCATM; BCT2; ECA40; mBcat; PP18

View more View less

UniProt ID: (Human) O15382, (Mouse) O35855, (Rat) O35854

View more View less

Entrez Gene ID: (Human) 587, (Mouse) 12036, (Rat) 64203

View more View less

Function(s)
branched-chain-amino-acid transaminase activity pyridoxal phosphate binding L-leucine transaminase activity L-valine transaminase activity L-isoleucine transaminase activity catalytic activity transaminase activity transferase activity
Process(es)
aspartate biosynthetic process branched-chain amino acid biosynthetic process branched-chain amino acid catabolic process leucine biosynthetic process valine biosynthetic process isoleucine metabolic process isoleucine catabolic process leucine metabolic process valine metabolic process metabolic process cellular amino acid biosynthetic process branched-chain amino acid metabolic process regulation of hormone levels lactation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0