Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Ataxin 2 Antibodies

Proteintech

Ataxin 2 Polyclonal Antibody, CoraLite® 594

View all (16) Ataxin 2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Ataxin 2 Polyclonal Antibody, CoraLite® 594

  • Antibody Testing Data (2)
Ataxin 2 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
Group 53 Created with Sketch.
Ataxin 2 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Ataxin 2 Antibody (CL594-21776) in IHC (PFA)

Immunofluorescent analysis of (4% PFA) fixed mouse brain tissue using CoraLite®594 Ataxin 2 antibody (CL594-21776) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Ataxin 2 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
Ataxin 2 Antibody in Flow Cytometry (Flow)
Ataxin 2 Polyclonal Antibody, CoraLite® 594

Product Details

CL594-21776

Applications
Tested Dilution
Publications

Immunohistochemistry (PFA fixed) (IHC (PFA))

1:50-1:500
-

Flow Cytometry (Flow)

0.4 µg/1x10^6 cell suspension
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant protein
View immunogen

Conjugate

CoraLite® 594 CoraLite® 594 CoraLite® 594

Excitation/Emission Max

593/614 nm

Form

Liquid

Purification

Antigen affinity chromatography

Storage buffer

PBS with 0.5% BSA, 50% glycerol

Contains

0.05% ProClin 300

Storage conditions

-20°C, store in dark

Shipping conditions

Wet ice

Product Specific Information

Immunogen sequence: GSDQRVVNGG VPWPSPCPSP SSRPPSRYQS GPNSLPPRAA TPTRPPSRPP SRPSRPPSHP SAHGSPAPVS TMPKRMSSEG PPRMSPKAQR HPRNHRVSAG RGSISSGLEF VSHNPPSEAA TPPVARTSPS GGTWSSVVSG VPRLSPKTHR PRSPRQNSIG NTPSGPVLAS PQAGIIPTEA VAMPIPAASP TPASPASNRA VTPSSEAKDS RLQDQRQNSP AGNKENIKPN ETSPSFSKAE NKGISPVVSE HRKQIDDLKK FKNDFRLQPS STSESMDQLL NKNREGEKSR DLIKDKIEPS AKDSFIENSS SNCTSGSSKP NSPSISPSIL SNTEHKRGPE VTSQGVQTSS

Target Information

The autosomal dominant cerebellar ataxias (ADCA) are a heterogeneous group of neurodegenerative disorders characterized by progressive degeneration of the cerebellum, brain stem and spinal cord. Clinically, ADCA has been divided into three groups: ADCA types I-III. Defects in this gene are the cause of spinocerebellar ataxia type 2 (SCA2). SCA2 belongs to the autosomal dominant cerebellar ataxias type I (ADCA I) which are characterized by cerebellar ataxia in combination with additional clinical features like optic atrophy, ophthalmoplegia, bulbar and extrapyramidal signs, peripheral neuropathy and dementia. SCA2 is caused by expansion of a CAG repeat in the coding region of this gene. Longer expansions result in earlier onset of the disease. Alternatively spliced transcript variants encoding different isoforms have been identified but their full length sequence has not been determined.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Ataxin-2; FLJ46772; spinocerebellar ataxia 2 (olivopontocerebellar ataxia 2, autosomal dominant, ataxin 2); spinocerebellar ataxia 2 homolog; Spinocerebellar ataxia type 2 protein; Spinocerebellar ataxia type 2 protein homolog; Trinucleotide repeat-containing gene 13 protein

View more View less

Gene Aliases: 9630045M23Rik; ASL13; ATX2; ATXN2; AW544490; SCA2; TNRC13

View more View less

UniProt ID: (Human) Q99700, (Mouse) O70305

View more View less

Entrez Gene ID: (Human) 6311, (Rat) 288663, (Mouse) 20239

View more View less

Function(s)
RNA binding epidermal growth factor receptor binding protein binding protein C-terminus binding poly(A) RNA binding molecular_function DBD domain binding
Process(es)
negative regulation of receptor internalization biological_process cerebellar Purkinje cell differentiation cytoplasmic mRNA processing body assembly stress granule assembly negative regulation of multicellular organism growth neuron projection morphogenesis homeostasis of number of cells neuromuscular process regulation of translation regulation of cytoplasmic mRNA processing body assembly RNA metabolic process RNA transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0