Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Artemin Antibodies

Invitrogen

Artemin Polyclonal Antibody, PeproTech®

View all (15) Artemin antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Artemin Polyclonal Antibody, PeproTech®

  • Antibody Testing Data (3)
Artemin Antibody in Western Blot (WB)
Group 53 Created with Sketch.
Artemin Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Artemin Antibody (500-P245-1MG) in WB

Western Blot: To detect Human Artemin by Western Blot analysis Artemin Polyclonal Antibody (Product # 500-P245-1MG) can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Artemin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Artemin Antibody in Western Blot (WB)
Artemin Antibody in Western Blot (WB)
Artemin Antibody in ELISA (ELISA)

Product Details

500-P245-1MG

Applications
Tested Dilution
Publications

Western Blot (WB)

0.1-0.2 µg/mL
-

ELISA (ELISA)

0.5-2.0 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit

Class

Polyclonal

Type

Antibody

Immunogen

E.coli-derived Recombinant Human Artemin

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

0.1-1.0 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS

Contains

no preservative

Storage conditions

-20°C

Shipping conditions

Ambient

RRID

AB_2930027

Product Specific Information

AA Sequence of recombinant protein: AGGPGSRARA AGARGCRLRS QLVPVRALGL GHRSDELVRF RFCSGSCRRA RSPHDLSLAS LLGAGALRPP PGSRPVSQPC CRPTRYEAVS FMDVNSTWRT VDRLSATACG CLG.

Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Artemin. Anti-Human Artemin-specific antibody was purified by affinity chromatography employing an immobilized Human Artemin matrix.

Sandwich ELISA: To detect Human Artemin by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5-2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human Artemin (500-P245Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Artemin.

Western Blot: To detect Human Artemin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Artemin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Target Information

The protein encoded by this gene is a member of the glial cell line-derived neurotophic factor (GDNF) family of ligands which are a group of ligands within the TGF-beta superfamily of signaling molecules. GDNFs are unique in having neurotrophic properties and have potential use for gene therapy in neurodegenerative disease. Artemin has been shown in culture to support the survival of a number of peripheral neuron populations and at least one population of dopaminergic CNS neurons. Its role in the PNS and CNS is further substantiated by its expression pattern in the proximity of these neurons. This protein is a ligand for the RET receptor and uses GFR-alpha 3 as a coreceptor. Multiple transcript variants encoding different isoforms have been found for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Artemin; Enovin; Neublastin; neurotrophic factor

View more View less

Gene Aliases: ARTN; ENOVIN; EVN; NBN

View more View less

UniProt ID: (Human) Q5T4W7

View more View less

Entrez Gene ID: (Human) 9048

View more View less

Function(s)
Ras guanyl-nucleotide exchange factor activity receptor binding growth factor activity
Process(es)
MAPK cascade signal transduction neuroblast proliferation axon guidance peripheral nervous system development positive regulation of GTPase activity induction of positive chemotaxis Peyer's patch morphogenesis lymphocyte migration into lymphoid organs
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-657bd577ff-sg76p:80/100.66.128.142:80.
git-commit: cdba60da2f3289684898c31dc50804c008488b19
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0