Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Apolipoprotein H Antibodies

Proteintech

Apolipoprotein H Monoclonal Antibody (4D9A4), CoraLite® Plus 488

View all (37) Apolipoprotein H antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Apolipoprotein H Monoclonal Antibody (4D9A4), CoraLite® Plus 488

  • Antibody Testing Data (1)
Apolipoprotein H Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Apolipoprotein H Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Apolipoprotein H Antibody (CL48866074100UL) in IHC (P)

Immunofluorescent analysis of (4% PFA) fixed human liver cancer tissue using CoraLite 488 Apolipoprotein H antibody (Product # CL48866074100UL), Clone: 4D9A4 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Apolipoprotein H Antibody in Immunohistochemistry (Paraffin) (IHC (P))

Product Details

CL48866074100UL

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1:200
-

Immunocytochemistry (ICC/IF)

1:50-1:500
-
Product Specifications

Species Reactivity

Human, Mouse, Pig, Rat

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

4D9A4

Conjugate

CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
  • Unconjugated
  • CoraLite 594
  • Request custom conjugation

Excitation/Emission Max

493/522 nm

Form

Liquid

Concentration

1 mg/mL

Purification

Protein G

Storage buffer

PBS, pH 7.3, with 50% glycerol, 0.5% BSA

Contains

0.05% ProClin 300

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles, store in dark

Shipping conditions

Wet ice

Target Information

Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 2GPI; Activated protein C-binding protein; Anticardiolipin cofactor; APC inhibitor; Apo-H; Apolipoprotein H; apolipoprotein H (beta-2-glycoprotein I); B2GPI; beta(2)GPI; Beta-2-glycoprotein 1; Beta-2-glycoprotein I

View more View less

Gene Aliases: APOH; B2G1; B2GP1; B2GPI; beta-2-GPI; beta2-GPI; BG

View more View less

UniProt ID: (Human) P02749, (Mouse) Q01339

View more View less

Entrez Gene ID: (Human) 350, (Rat) 287774, (Mouse) 11818

View more View less

Function(s)
glycoprotein binding protein binding phospholipid binding heparin binding lipid binding identical protein binding lipoprotein lipase activator activity
Process(es)
negative regulation of endothelial cell proliferation triglyceride metabolic process blood coagulation blood coagulation, intrinsic pathway negative regulation of endothelial cell migration negative regulation of angiogenesis regulation of blood coagulation negative regulation of blood coagulation organ regeneration plasminogen activation negative regulation of myeloid cell apoptotic process triglyceride transport negative regulation of smooth muscle cell apoptotic process positive regulation of lipoprotein lipase activity regulation of fibrinolysis negative regulation of fibrinolysis platelet degranulation aging positive regulation of triglyceride catabolic process positive regulation of blood coagulation response to triglyceride negative regulation of respiratory burst
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0